Skip to Content
Merck
All Photos(1)

Documents

408298

Sigma-Aldrich

Di(ethylene glycol) ethyl ether acrylate

technical grade, contains 1000 ppm monomethyl ether hydroquinone as inhibitor

Synonym(s):

Carbitol acrylate, 2-(2-Ethoxyethoxy)ethyl acrylate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
H2C=CHCO2(CH2CH2O)2C2H5
CAS Number:
Molecular Weight:
188.22
EC Number:
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:
NACRES:
NA.23

grade

technical grade

Quality Level

Assay

≥ 85.0%

form

liquid

contains

1000 ppm monomethyl ether hydroquinone as inhibitor

refractive index

n20/D 1.439 (lit.)

bp

95 °C/5 mmHg (lit.)

density

1.016 g/mL at 25 °C (lit.)

SMILES string

CCOCCOCCOC(=O)C=C

InChI

1S/C9H16O4/c1-3-9(10)13-8-7-12-6-5-11-4-2/h3H,1,4-8H2,2H3

InChI key

FTALTLPZDVFJSS-UHFFFAOYSA-N

Pictograms

Skull and crossbonesEnvironment

Signal Word

Danger

Hazard Classifications

Acute Tox. 3 Dermal - Acute Tox. 4 Oral - Aquatic Chronic 2 - Eye Irrit. 2 - Skin Irrit. 2 - Skin Sens. 1A

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Kazuhiro Sato et al.
Macromolecular bioscience, 15(9), 1296-1303 (2015-05-29)
Six types of poly(2-methoxyethyl acrylate) (PMEA) analogues were synthesized and the water structure in the hydrated polymers was characterized using differential scanning calorimetry (DSC). The hydrated PMEA analogues exhibited the different amounts of intermediate water. Non-thrombogenicity evaluation was performed on
Steven Martens et al.
Nature communications, 9(1), 4451-4451 (2018-10-28)
Sequence-defined macromolecules consist of a defined chain length (single mass), end-groups, composition and topology and prove promising in application fields such as anti-counterfeiting, biological mimicking and data storage. Here we show the potential use of multifunctional sequence-defined macromolecules as a

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service