Skip to Content
Merck
All Photos(1)

Documents

345237

Sigma-Aldrich

[1,2-Bis(diphenylphosphino)ethane]dichloropalladium(II)

98%

Synonym(s):

PdCl2(dppe)

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
[(C6H5)2PCH2CH2P(C6H5)2]PdCl2
CAS Number:
Molecular Weight:
575.74
MDL number:
UNSPSC Code:
12161600
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

powder

reaction suitability

reaction type: Buchwald-Hartwig Cross Coupling Reaction
reaction type: Cross Couplings
reaction type: Heck Reaction
reaction type: Hiyama Coupling
reaction type: Negishi Coupling
reaction type: Sonogashira Coupling
reaction type: Stille Coupling
reaction type: Suzuki-Miyaura Coupling
reagent type: catalyst
core: palladium

SMILES string

Cl[Pd]Cl.C(CP(c1ccccc1)c2ccccc2)P(c3ccccc3)c4ccccc4

InChI

1S/C26H24P2.2ClH.Pd/c1-5-13-23(14-6-1)27(24-15-7-2-8-16-24)21-22-28(25-17-9-3-10-18-25)26-19-11-4-12-20-26;;;/h1-20H,21-22H2;2*1H;/q;;;+2/p-2

InChI key

LDJXFZUGZASGIW-UHFFFAOYSA-L

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Leila Tabrizi et al.
Dalton transactions (Cambridge, England : 2003), 46(41), 14164-14173 (2017-10-07)
New multinuclear gold(iii), palladium(ii) pincer complexes containing bis(diphenylphosphino) ferrocene/non-ferrocene ligands of formula [(L)Au(μ2-η2-CS3)Pd(dppf)](PF6)2, 1, and [(L)Au(μ2-η2-CS3)Pd(dppe)](PF6)2, 2 (HL = 5-methoxy-1,3-bis (1-methyl-1H-benzo[d]imidazol-2-yl)benzene, dppf = 1,1'-bis(diphenylphosphino)ferrocene, and dppe = bis(diphenylphosphino)ethane) have been synthesized and fully characterized. Both complexes are more cytotoxic to

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service