Skip to Content
Merck
All Photos(1)

Key Documents

331066

Sigma-Aldrich

1-Phenylnonane

96%

Synonym(s):

Nonylbenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5(CH2)8CH3
CAS Number:
Molecular Weight:
204.35
Beilstein:
1907173
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

96%

form

liquid

refractive index

n20/D 1.483 (lit.)

bp

282 °C (lit.)

density

0.858 g/mL at 25 °C (lit.)

SMILES string

CCCCCCCCCc1ccccc1

InChI

1S/C15H24/c1-2-3-4-5-6-7-9-12-15-13-10-8-11-14-15/h8,10-11,13-14H,2-7,9,12H2,1H3

InChI key

LIXVMPBOGDCSRM-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

1-Phenylnonane was employed as a solvent:
  • in high-yielding synthesis of meso-tetraalkylporphyrins
  • to examine the 2D structures by changing the alkyl chain length of the isobutenyl compounds

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Rémi Plamont et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 19(34), 11293-11300 (2013-07-11)
We have developed a high-yielding synthesis of meso-tetraalkylporphyrins, which previously have been obtained only in lower yields. By employing Montmorrilonite K10 as the acid catalyst and 3 Å molecular sieves as the dehydrating agent, yields that reached 70 % could be achieved
Yoshihiro Kikkawa et al.
Organic & biomolecular chemistry, 10(40), 8087-8094 (2012-09-12)
A series of isobutenyl compounds possessing various alkyl chain lengths (C(n)-1) with a carbon number of n = 14-21 were synthesized and their two-dimensional (2D) structures were systematically studied using scanning tunnelling microscopy (STM) at a highly oriented pyrolytic graphite

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service