Skip to Content
Merck
All Photos(1)

Key Documents

291110

Sigma-Aldrich

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane

98%

Synonym(s):

Cryptand 222, Kryptofix® 222

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C18H36N2O6
CAS Number:
Molecular Weight:
376.49
Beilstein:
620282
EC Number:
MDL number:
UNSPSC Code:
12352005
PubChem Substance ID:
NACRES:
NA.22

Assay

98%

form

solid

mp

68-71 °C (lit.)

SMILES string

C1COCCN2CCOCCOCCN(CCO1)CCOCCOCC2

InChI

1S/C18H36N2O6/c1-7-21-13-14-24-10-4-20-5-11-25-17-15-22-8-2-19(1)3-9-23-16-18-26-12-6-20/h1-18H2

InChI key

AUFVJZSDSXXFOI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane(cryptand 222, Kryptofix 222) is an organic compound with formula C18H36N2O6. This compound has a cage-like three-dimensional structure that donates N2O6. The ligand has shown a wide range of uses in magnetic resonance imaging, organic synthesis, crystallography, electrochemistry, and chromatography. In solution, it is a well-known sequestering agent for metal ions.

Application

4,7,13,16,21,24-Hexaoxa-1,10-diazabicyclo[8.8.8]hexacosane is used as:

  • A electrolyte additive in isotachophoresis[separation method that is particularly suited to the analysis of small ions]for the analysis of alkali metal cations.
  • A complexing agent in the preparation of K+-imprinted nanoparticles using methacrylic acid as the functional monomer, ethylene glycol dimethacrylate as the crosslinker and 2,2′-azobisisobutyronitrile as the radical initiator.
  • A structure directing agent (SDA) in the preparation of LTA-type AlPO4 crystals.

Packaging

Bottomless glass bottle. Contents are inside inserted fused cone.
Cryptand used with potassium mirror to reduce a hindered distannene to a crystalline radical anion.

Legal Information

Kryptofix is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Metal Ion Compiexing by Cryptand 222 in Solutions. A Thermodynamic Approach
Marcus Y
Reviews in Analytical Chemistry, 23, 269-302 (2004)
Preparation of large and well-shaped LTA-type AlPO4 crystals by using crown ether Kryptofix 222 as structure directing agent
Huang A, et al.
Microporous and Mesoporous Materials : The Official Journal of the International Zeolite Association, 129, 90-99 (2010)
Dynamers at the solid-liquid interface: controlling the reversible assembly/reassembly process between two highly ordered supramolecular guanine motifs.
Artur Ciesielski et al.
Angewandte Chemie (International ed. in English), 49(11), 1963-1966 (2010-02-06)
Direct analysis of Kryptofix 2.2.2 in 18FDG by gas chromatography using a nitrogen-selective detector.
R A Ferrieri et al.
Nuclear medicine and biology, 20(3), 367-369 (1993-04-01)
M Castaing et al.
The Journal of membrane biology, 89(3), 251-267 (1986-01-01)
The kinetics of K+ and Na+ transport across the membrane of large unilamellar vesicles (L.U.V.) were compared at two pH's, with two carriers: (222)C10-cryptand (diaza-1,10-decyl-5-hexaoxa-4,7,13,16,21,24-bicyclo[8.8.8.]+ ++hexacosane) and valinomcyin, i.e. an ionizable macrobicyclic amino polyether and a neutral macrocyclic antibiotic. The

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service