All Photos(1)
About This Item
Linear Formula:
CF3C6H4CH2NH2
CAS Number:
Molecular Weight:
175.15
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
Assay
98%
refractive index
n20/D 1.463 (lit.)
density
1.222 g/mL at 25 °C (lit.)
functional group
amine
fluoro
SMILES string
NCc1cccc(c1)C(F)(F)F
InChI
1S/C8H8F3N/c9-8(10,11)7-3-1-2-6(4-7)5-12/h1-4H,5,12H2
InChI key
YKNZTUQUXUXTLE-UHFFFAOYSA-N
Related Categories
Application
3-(Trifluoromethyl)benzylamine has been used in the preparation of 6-substituted purines.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Skin Corr. 1B
Storage Class Code
8A - Combustible corrosive hazardous materials
WGK
WGK 2
Flash Point(F)
159.8 °F - closed cup
Flash Point(C)
71 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Synthesis of a new series of purine derivatives and their anti-cyclin-dependent kinase activities.
Legraverend M, et al.
Journal of Heterocyclic Chemistry, 38(1), 299-303 (2001)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service