Skip to Content
Merck
All Photos(1)

Key Documents

247383

Sigma-Aldrich

Boron tribromide dimethyl sulfide complex solution

1.0 M in methylene chloride

Synonym(s):

Dimethyl sulfide-tribromoborane, Tribromoborane-methyl sulfide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)2S·BBr3
CAS Number:
Molecular Weight:
312.66
MDL number:
UNSPSC Code:
12352101
PubChem Substance ID:
NACRES:
NA.22

form

liquid

concentration

1.0 M in methylene chloride

density

1.456 g/mL at 25 °C

functional group

thioether

SMILES string

C[S+](C)[B-](Br)(Br)Br

InChI

1S/C2H6BBr3S/c1-7(2)3(4,5)6/h1-2H3

InChI key

NCVLHAUANAMSTL-UHFFFAOYSA-N

Application

Boron tribromide dimethyl sulfide complex (BBr3S(CH3)2) can be used as a reagent:
  • To prepare demethylated products from aryl methyl ethers by deprotection of methyl groups.
  • To hydrolyze 1,3-methylenedioxole ring compounds to 1,2-dihydroxy compounds.
  • To prepare benzyl S,S′-diphenyl acetal by reacting with phenylacetic acid and thexylphenylthioborane.
  • To synthesize Baylis−Hillman adducts and α-halomethyl enones by the reaction between aldehydes and 3-buten-2-one.

Reactant for preparation of:
  • Precursor of HIV protease inhibitor KNI 764 using cation-exchange resin mediated Mannich reaction

Reagent for:
  • Baylis-Hillman reactions

Signal Word

Danger

Hazard Statements

Hazard Classifications

Carc. 2 - Eye Dam. 1 - Skin Corr. 1B - STOT SE 3

Target Organs

Central nervous system

Supplementary Hazards

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Boron trihalide-methyl sulfide complexes as convenient reagents for dealkylation of aryl ethers
Williard PG and Fryhle CB
Tetrahedron Letters, 21(39), 3731-3734 (1980)
Tetrahedron Letters, 21, 3731-3731 (1980)
Dimethyl sulfide-boron trihalide-mediated reactions of α, β, unsaturated ketones with aldehydes: one-pot synthesis of Baylis-Hillman adducts and ?-halomethyl enones
Iwamura T, et al.
Tetrahedron, 57(40), 8455-8462 (2001)
Direct conversion of carboxylic acids and carboxylic esters into S, S?-diphenyl acetals and phenyl sulfides with thexylphenylthioborane
Kim S and Kim SS
Tetrahedron Letters, 28(17), 1913-1916 (1987)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service