Skip to Content
Merck
All Photos(1)

Documents

197734

Sigma-Aldrich

Trimethyl(phenyl)silane

99%

Synonym(s):

(Trimethylsilyl)benzene, Phenyltrimethylsilane

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5Si(CH3)3
CAS Number:
Molecular Weight:
150.29
Beilstein:
1855012
EC Number:
MDL number:
UNSPSC Code:
12352103
PubChem Substance ID:
NACRES:
NA.22

Quality Level

Assay

99%

form

liquid

refractive index

n20/D 1.491 (lit.)

bp

168-170 °C (lit.)

density

0.873 g/mL at 25 °C (lit.)

SMILES string

C[Si](C)(C)c1ccccc1

InChI

1S/C9H14Si/c1-10(2,3)9-7-5-4-6-8-9/h4-8H,1-3H3

InChI key

KXFSUVJPEQYUGN-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameCorrosion

Signal Word

Danger

Hazard Statements

Precautionary Statements

Hazard Classifications

Aquatic Chronic 4 - Eye Dam. 1 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

104.0 °F - closed cup

Flash Point(C)

40 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Gregory J Anthony et al.
Medical physics, 46(2), 774-788 (2018-11-11)
In this study, the efficacy of transurethral prostate ablation in the presence of silica-shell ultrasound-triggered phase-shift emulsions (sUPEs) doped with MR contrast was evaluated. The influence of sUPEs on MR imaging assessment of the ablation zone was also investigated. sUPEs

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service