All Photos(3)
About This Item
Linear Formula:
HCOC6H4CH=CHCO2H
CAS Number:
Molecular Weight:
176.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Recommended Products
form
solid
mp
251-254 °C (dec.) (lit.)
functional group
aldehyde
carboxylic acid
SMILES string
OC(=O)\C=C\c1ccc(C=O)cc1
InChI
1S/C10H8O3/c11-7-9-3-1-8(2-4-9)5-6-10(12)13/h1-7H,(H,12,13)/b6-5+
InChI key
LBOUHDMYVURTMA-AATRIKPKSA-N
Application
4-Formylcinnamic acid may be used in the synthesis of (E)-4-[(5,6,7,8-tetrahydro-5,5,8,8-tetramethylnaphth-2-yl)-ethynyl]-cinnamic acid.
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service