Merck
  • Bovicin HJ50-like lantibiotics, a novel subgroup of lantibiotics featured by an indispensable disulfide bridge.
Bovicin HJ50-like lantibiotics, a novel subgroup of lantibiotics featured by an indispensable disulfide bridge.
PloS one (2014-05-14)
Jian Wang, Hongchu Ma, Xiaoxuan Ge, Jie Zhang, Kunling Teng, Zhizeng Sun, Jin Zhong
ABSTRACT
Lantibiotics are ribosomally-synthesized and posttranslationally modified peptides with potent antimicrobial activities. Discovery of novel lantibiotics has been greatly accelerated with the soaring release of genomic information of microorganisms. As a unique class II lantibiotic, bovicin HJ50 is produced by Streptococcus bovis HJ50 and contains one rare disulfide bridge. By using its precursor BovA as a drive sequence, 16 BovA-like peptides were revealed in a wide variety of species. From them, three representative novel lan loci from Clostridium perfringens D str. JGS1721, Bacillus cereus As 1.348 and B. thuringiensis As 1.013 were identified by PCR screening. The corresponding mature lantibiotics designated perecin, cerecin and thuricin were obtained and structurally elucidated to be bovicin HJ50-like lantibiotics especially by containing a conserved disulfide bridge. The disulfide bridge was substantiated to be essential for the function of bovicin HJ50-like lantibiotics as its disruption eliminated their antimicrobial activities. Further analysis indicated that the disulfide bridge played a crucial role in maintaining the hydrophobicity of bovicin HJ50, which might facilitate it to exert antimicrobial function. This study unveiled a novel subgroup of disulfide-containing lantibiotics from bacteria of different niches and further demonstrated the indispensable role of disulfide bridge in these novel bovicin HJ50-like lantibiotics.
Sigma-Aldrich
DL-Dithiothreitol, BioUltra, for molecular biology, ≥99.5% (RT)
Sigma-Aldrich
DL-Dithiothreitol, ≥99.0% (RT)
Sigma-Aldrich
DL-Dithiothreitol, ≥98% (HPLC), ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, suitable for electrophoresis, ≥99%
Sigma-Aldrich
DL-Dithiothreitol, for molecular biology, ≥98% (HPLC), ≥99% (titration)
Sigma-Aldrich
DL-Dithiothreitol, BioXtra, ≥99.0% (titration)
Sigma-Aldrich
DL-Dithiothreitol, Vetec, reagent grade, 98%
Sigma-Aldrich
N-Ethylmaleimide, purum p.a., ≥99.0% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioUltra, ≥99.0% (HPLC)
Sigma-Aldrich
Phosphorus, red, ≥99.99% trace metals basis
Sigma-Aldrich
Phosphorus, red, ≥97.0%
Sigma-Aldrich
Bis(1,3-dibutylbarbituric acid) trimethine oxonol, ≥95% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, BioXtra, ≥98% (HPLC)
Sigma-Aldrich
N-Ethylmaleimide, crystalline, ≥98% (HPLC)