Skip to Content
Merck
All Photos(2)

Documents

V900830

Sigma-Aldrich

DL-Dithiothreitol

Vetec, reagent grade, 98%

Synonym(s):

threo-1,4-Dimercapto-2,3-butanediol, Cleland’s reagent, DTT

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
HSCH2CH(OH)CH(OH)CH2SH
CAS Number:
Molecular Weight:
154.25
Beilstein:
1719757
EC Number:
MDL number:
UNSPSC Code:
12352209
PubChem Substance ID:

grade

reagent grade

product line

Vetec

Assay

98%

mp

41-44 °C (lit.)

storage temp.

2-8°C

SMILES string

O[C@H](CS)[C@H](O)CS

InChI

1S/C4H10O2S2/c5-3(1-7)4(6)2-8/h3-8H,1-2H2/t3-,4-/m1/s1

InChI key

VHJLVAABSRFDPM-QWWZWVQMSA-N

Looking for similar products? Visit Product Comparison Guide

Application

An excellent reagent for maintaining SH groups in reduced state; quantitatively reduces disulfides. DTT is effective in sample buffers for reducing protein disulfide bonds prior to SDS-PAGE. DTT can also be used for reducing the disulfide bridge of the cross-linker N,N′-bis(acryloyl)cystamine to break apart the matrix of a polyacrylamide gel. DTT is less pungent and is less toxic than 2-mercaptoethanol. Typically, a seven fold lower concentration of DTT (100 mM) is needed than is used for 2-mercaptoethanol (5% v/v, 700 mM).

Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

CorrosionExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Dam. 1 - Skin Irrit. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Qingfang Li et al.
Frontiers in oncology, 9, 1489-1489 (2020-01-31)
Objective: Extranodal natural killer /T-cell lymphoma (ENKTL) is an aggressive and unusual subtype of non-Hodgkin lymphoma (NHL) that it is related with the Epstein-Barr virus (EBV). CSF is considered as an ideal source of high-concenrtation disease-related proteins. We aimed at
A A Vorster et al.
Clinical genetics, 87(5), 492-495 (2014-04-29)
Spondyloepimetaphyseal dysplasia with joint laxity (SEMD-JL), type 1 is an autosomal recessive disorder which has been identified in more than 30 affected children in the Afrikaans-speaking community of South Africa. Sequencing of B3GALT6 revealed a specific mutation, c.235A > G, in homozygous
Mojtaba Abdul Roda et al.
PloS one, 9(5), e97594-e97594 (2014-05-20)
A novel neutrophil chemoattractant derived from collagen, proline-glycine-proline (PGP), has been recently characterized in chronic obstructive pulmonary disease (COPD). This peptide is derived via the proteolytic activity of matrix metalloproteases (MMP's)-8/9 and PE, enzymes produced by neutrophils and present in
Jian Wang et al.
PloS one, 9(5), e97121-e97121 (2014-05-14)
Lantibiotics are ribosomally-synthesized and posttranslationally modified peptides with potent antimicrobial activities. Discovery of novel lantibiotics has been greatly accelerated with the soaring release of genomic information of microorganisms. As a unique class II lantibiotic, bovicin HJ50 is produced by Streptococcus
Maria Gschwandtner et al.
The Journal of investigative dermatology, 134(8), 2192-2201 (2014-04-04)
Antimicrobial peptides (AMPs), an important part of the innate immune system, are crucial for defense against invading microorganisms. Whereas AMPs have been extensively studied in adult skin, little is known about the impact of AMPs in the developing human skin.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service