Merck
  • Palladium-catalyzed direct arylation of nitro-substituted aromatics with aryl halides.
Palladium-catalyzed direct arylation of nitro-substituted aromatics with aryl halides.
Organic letters (2008-09-25)
Laurence Caron, Louis-Charles Campeau, Keith Fagnou
ABSTRACT
Direct arylation reactions of nitrobenzenes and aryl halides occur in good yield and high ortho regioselectivity. These reactions can be performed on gram scale with as few as 3 equiv of the nitro arene relative to the aryl halide. The synthetic utility of this method is demonstrated via rapid synthesis of a Boscalid intermediate.
Sigma-Aldrich
Di-tert-butyl(methyl)phosphonium tetrafluoroborate, 97%