Merck
Catalytic intermolecular direct arylation of perfluorobenzenes.
Journal of the American Chemical Society (2006-07-06)
Marc Lafrance, Christopher N Rowley, Tom K Woo, Keith Fagnou
ABSTRACT
Penta-, tetra-, tri-, and difluorobenzenes undergo direct arylation with a wide range of arylhalides in high yield. Inverse reactivity is observed compared to the common electrophilic aromatic substitution pathway since electron-deficient, C-H acidic arenes react preferentially. Computational studies indicate that C-H bond cleavage occurs via a concerted carbon-palladium and carbon-hydrogen bond cleaving event involving a carbonate or a bromide ligand. The reactions are rapid, require only a slight excess of the perfluoroarene reagent, and utilize commercially available, air-stable catalyst precursors.
Sigma-Aldrich
Di-tert-butyl(methyl)phosphonium tetrafluoroborate, 97%