![Di-tert-butyl(methyl)phosphonium tetrafluoroborate 97%](/deepweb/assets/sigmaaldrich/product/structures/299/679/0440f34c-c7c0-47bc-b015-54ef87182843/640/0440f34c-c7c0-47bc-b015-54ef87182843.png)
Di-tert-butyl(methyl)phosphonium tetrafluoroborate
![Di-tert-butyl(methyl)phosphonium tetrafluoroborate 97%](/deepweb/assets/sigmaaldrich/product/structures/299/679/0440f34c-c7c0-47bc-b015-54ef87182843/640/0440f34c-c7c0-47bc-b015-54ef87182843.png)
Synonym(s):
(t-Bu)2PMeHBF4, Bis(1,1-dimethylethyl)methylphosphine tetrafluoroborate
Empirical Formula (Hill Notation):
C9H22BF4P
CAS Number:
Molecular Weight:
248.05
Products
Brand
Formula Weight
Melting Point (°C)
pH Value
Physical Form
Purity
Quality Segment
Functional Group
Reaction Type
Reagent Type
Showing 1-1 of 1
Compare
Product Number
Description
Pricing