1171455
USP
Dermatan sulfate
United States Pharmacopeia (USP) Reference Standard
Synonym(s):
Chondroitin sulfate B sodium salt, β-Heparin, Dermatan sulfate sodium salt
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
grade
pharmaceutical primary standard
API family
dermatan sulfate
manufacturer/tradename
USP
application(s)
pharmaceutical (small molecule)
format
neat
storage temp.
2-8°C
Looking for similar products? Visit Product Comparison Guide
General description
Dermatan sulfate is a glycosaminoglycan (GAG) that is mainly located in the skin and is also found in various mammalian tissues. It is made up of repeating units of N-acetyl-d-galactosamine (GalNAc) and l-iduronic acid (IdoA). Dermatan sulfate interacts with proteoglycans and plays a role in regulating several biological activities, including coagulation, angiogenesis, tumor migration, and growth factor expression.
Dermatan sulfate USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia
Dermatan sulfate USP reference standard is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets, has been developed and issued under the authority of the issuing Pharmacopoeia. For further information and support please go to the website of the issuing Pharmacopoeia
Application
The Dermatan sulfate USP reference standard is intended for use in specified quality tests and assays. It is also used to prepare system suitability solution for chromatographic identity using liquid chromatography coupled to a UV detector according to the monograph of United States Pharmacopeia (USP).
Further information is available in the monograph, Heparin Sodium, USP43-NF38 - 2195 of the USP (United States Pharmacopeia).
Further information is available in the monograph, Heparin Sodium, USP43-NF38 - 2195 of the USP (United States Pharmacopeia).
Other Notes
This product is part of the USP Biologics program.
Sales restrictions may apply.
related product
Product No.
Description
Pricing
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Heparin Sodium
United States Pharmacopeia, 41(6) (2018)
Modular automated solid phase synthesis of dermatan sulfate oligosaccharides
Chemical Communications (Cambridge, England) (2014)
Heparin Sodium
United States Pharmacopeia, 41(6), 2195-2195 (2018)
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service