Skip to Content
Merck
All Photos(1)

Documents

1009901

USP

Acetyltributyl citrate

United States Pharmacopeia (USP) Reference Standard

Synonym(s):

Tributyl 2-acetylcitrate, Acetyl tributyl citrate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CO2C[CO2(CH2)3CH3][CH2CO2(CH2)3CH3]2
CAS Number:
Molecular Weight:
402.48
Beilstein:
2303316
MDL number:
UNSPSC Code:
41116107
PubChem Substance ID:
NACRES:
NA.24

grade

pharmaceutical primary standard

vapor pressure

0.26 psi ( 20 °C)
0.8 psi ( 55 °C)

description

API family: acetyltributyl citrate

API family

acetyltributyl citrate

manufacturer/tradename

USP

refractive index

n20/D 1.443 (lit.)

density

1.05 g/mL at 25 °C (lit.)

application(s)

pharmaceutical (small molecule)

format

neat

SMILES string

CCCCOC(=O)CC(CC(=O)OCCCC)(OC(C)=O)C(=O)OCCCC

InChI

1S/C20H34O8/c1-5-8-11-25-17(22)14-20(28-16(4)21,19(24)27-13-10-7-3)15-18(23)26-12-9-6-2/h5-15H2,1-4H3

InChI key

QZCLKYGREBVARF-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

This product is provided as delivered and specified by the issuing Pharmacopoeia. All information provided in support of this product, including SDS and any product information leaflets have been developed and issued under the Authority of the issuing Pharmacopoeia.For further information and support please go to the website of the issuing Pharmacopoeia.

Application

Acetyltributyl citrate USP reference standard, intended for use in specified quality tests and assays as specified in the USP compendia. Also, for use with USP monographs such as:
  • Tributyl Citrate

Analysis Note

These products are for test and assay use only. They are not meant for administration to humans or animals and cannot be used to diagnose, treat, or cure diseases of any kind.  ​

Other Notes

Sales restrictions may apply.

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

422.6 °F - closed cup

Flash Point(C)

217 °C - closed cup


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service