Skip to Content
Merck
All Photos(1)

Key Documents

CRM16806

Supelco

Pymetrozin Metabolite CGA313124

certified reference material, TraceCERT®, Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

Synonym(s):

6-Hydroxymethyl-4-(pyridin-3-ylmethylenamino)-4,5-dihydro-1,2,4-triazin-3(2H)one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C10H11N5O2
Molecular Weight:
233.23
UNSPSC Code:
41116107
NACRES:
NA.24

grade

certified reference material
TraceCERT®

Quality Level

product line

TraceCERT®

shelf life

limited shelf life, expiry date on the label

manufacturer/tradename

Manufactured by: Sigma-Aldrich Production GmbH, Switzerland

format

neat

storage temp.

2-8°C

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This certified reference material (CRM) is produced and certified in accordance with ISO/IEC 17025 and ISO 17034. This CRM is traceable to primary material from an NMI, e.g. NIST or NMIJ.
Certified content by quantitative NMR incl. uncertainty and expiry date are given on the certificate.
Download your certificate at: http://www.sigma-aldrich.com

Pymetrozin is a synthetic herbicide belonging to the pyridine azomethrin family, used to control aphids, plant hoppers, and whiteflies in fruits, vegetables, flowers, and potatoes. Pymetrozin shows an immediate inhibition of feeding in aphids, causing death by starvation.

Pymetrozin has to be monitored in the Multiannual Control Programme for Pesticides Residues (MACP), run within the EU and EFTA in/on products of plant origin.

Pymetrozin is not approved for use in European Union as per Regulation (EC) No 1107/2009. Maximum residue levels (MRLs) for Pymetrozin have been set according to Regulation (EU) No 2021/155 from 0.01 to 0.1 mg/kg for various products of plant and animal origin.

Application

The certified reference material (CRM) is intended to be used as a calibrant for chromatography and other analytical techniques. The CRM may also find following uses:
  • To evaluate tetradifon, deltametrin, and pymetrozin pesticide residues on cucumbers by gas chromatography
  • To investigate pymetrozin residues in cucumber under field conditions
  • To assess the pesticide residues in strawberries subjected to various pesticide treatment regimes
  • To evaluate the side effects of hexaflumuron, pymetrozin, and spirodiclofen and some plant extracts on life table parameters of Chrysoperla carnea (Stephens) under controlled conditions
  • To evaluate different life table parameters as an indicator of the sub-lethal effects of four chemical pesticides (pymetrozin, diazinon, imidacloprid, and abamectin) and Calotropis procera extract on Bemisia tabaci

Legal Information

TraceCERT is a registered trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Health hazard

Signal Word

Warning

Hazard Statements

Hazard Classifications

Aquatic Chronic 3 - Carc. 2 - Repr. 2

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jana Kovacova et al.
Food additives & contaminants. Part A, Chemistry, analysis, control, exposure & risk assessment, 30(12), 2123-2135 (2013-12-07)
The dynamics of pesticide residues in strawberries that involved quantification of pesticide residues in ripe fruits after model treatment was evaluated in repeated field trials conducted over 3 years. Sixteen commercial pesticide formulations in various combinations were employed in applications
K Talebi et al.
Communications in agricultural and applied biological sciences, 71(2 Pt A), 75-78 (2007-03-30)
An experiment was carried out to investigate the residues of pymetrozin in cucumber under field conditions. Plots of cucumber (cv. Daminus) were sprayed with pymetrozine (Chess, WG50) at two different rates; 0.5 and 1.0 kg/ha. Samples were collected 2 hours
K Talebi et al.
Communications in agricultural and applied biological sciences, 71(2 Pt A), 75-78 (2007-03-30)
An experiment was carried out to investigate the residues of pymetrozin in cucumber under field conditions. Plots of cucumber (cv. Daminus) were sprayed with pymetrozine (Chess, WG50) at two different rates; 0.5 and 1.0 kg/ha. Samples were collected 2 hours

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service