Documents
58721
Tubing Adapter
1/4-28 male UNF, female Luer
About This Item
UNSPSC Code:
41121815
material
Kel-F (CTFE)
description
for chromatography applications
packaging
pkg of 1 ea
manufacturer/tradename
VICI Valco® CFLAPFA
fitting
female Luer
1/4-28 male UNF
General description
Female adapter (1/16" I.D.) connects syringe to ¼"-28 threaded tubing or adapter.
Tubing adaptor is a national pipe thread taper made up of stainless steel which is used to connect a female adaptor. It may be used to connect capillary tubes in HPLC, GC etc.
Application
Tubing adaptor was used in solid phase extraction.
Legal Information
Valco is a registered trademark of Valco Instruments Co. Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Sorry, we don't have COAs for this product available online at this time.
Visit the Document Library
Customers Also Viewed
Slide 1 of 2
1 of 2
C Brede et al.
Food additives and contaminants, 21(11), 1115-1124 (2005-03-15)
Migration of aniline from polyamide cooking utensils was investigated. Aniline was found to originate from the application of black colorant in the polyamide 66 raw material. Analysis of polyamide raw material revealed an aniline concentration of 121 +/- 13 mg