509396
SGE Gas Tight Syringe, Removable Needle
2.5MDR-GT, volume 2500 μL, needle size 23 ga, needle L × O.D. 50 mm × 0.63 mm
Synonym(s):
SYRINGE SGE 2.5ML, GAS TIGHT
All Photos(1)
About This Item
Recommended Products
material
bevel tip stainless steel needle (point style 2)
needle size
23 ga
feature
removable needle
packaging
pkg of 1 ea
manufacturer/tradename
SGE 008500
needle L × O.D.
50 mm × 0.63 mm
volume
2500 μL
General description
SGE gas tight syringe with removable needle, Model: 2.5MDR-GT (volume 2.5 mL, needle size 23 ga (bevel tip), needle L × O.D. 50 mm × 0.63 mm)
- Made of borosilicate glass, PTFE plunger tips, and KEL-F Luer cones.
- Autoclavable syringes but the plunger should be removed.
- Features an interference fit between the plunger tip and the barrel which enhances the gas sealing properties of the syringe.
- Maximum inertness for liquid or gas samples.
- Suitable for manual dispenser/diluter applications.
- High accuracy of dispensed volume.
Choose from one of the most recent versions:
Certificates of Analysis (COA)