Skip to Content
Merck
All Photos(1)

Documents

46985-U

Supelco

Non-volatile Acid Standard Mix

0.01 meq/mL each component in deionized water, analytical standard

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
85151701
NACRES:
NA.24

grade

analytical standard

Quality Level

CofA

current certificate can be downloaded

packaging

pkg of 100 mL

concentration

0.01 meq/mL each component in deionized water

technique(s)

HPLC: suitable
gas chromatography (GC): suitable

application(s)

environmental

format

multi-component solution

storage temp.

2-8°C

General description

Non-volatile Acid Standard Mix may be used for the qualitative determination of free fatty acids in aqueous solutions. It is recommended that it should be purchased from chemical manufacturers with knowledge in the preparation, handling, storage, and shipment of volatile analytes.

Application

Non-volatile Acid Standard Mix is used as an analytical standard in the thermal separation of organic carbon (OC) and elemental carbon (EC) for subsequent radiocarbon analysis.
Refer to the product′s Certificate of Analysis for more information on a suitable instrument technique. Contact Technical Service for further support.

Analyte

Description

    Fumaric acid
    DL-Lactic acid
    Malonic acid
    Methylmalonic acid
    Oxalacetic acid
    Oxalic acid
    Pyruvic acid
    Succinic acid
    See All (8)

Storage Class Code

12 - Non Combustible Liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Evaluation of a 2-step thermal method for separating organic and elemental carbon for radiocarbon analysis
Dusek U, et al.
Atmospheric Measurement Techniques Discussions, 7(1), 131-169 (2014)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service