Skip to Content
Merck
All Photos(2)

Key Documents

20556

Supelco

Glass Cartridge for Small PUF Plug

pkg of 1 ea

Synonym(s):

Small PUF Cartridges

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12190000

material

glass cartridge

packaging

pkg of 1 ea

technique(s)

active air sampling: suitable

application(s)

air monitoring
environmental
industrial hygiene

compatibility

for use with ORBO-1000 (22 mm PUF plug)

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service