Z740218
PTLC (Preparative TLC) Glass Plates
L × W 20 cm × 20 cm, silica gel 60 matrix, binder, Polymeric, fluorescent indicator
Synonym(s):
Preparative Thin-Layer Chromatography Glass Plates
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
glass support
silica gel 60 matrix
Quality Level
feature
binder Polymeric
fluorescent indicator
packaging
pkg of 20 ea
manufacturer/tradename
EMD Millipore® 1.05744.0001
technique(s)
thin layer chromatography (TLC): suitable
L × W
20 cm × 20 cm
layer thickness
500 μm
pore size
60 Å
Looking for similar products? Visit Product Comparison Guide
Legal Information
EMD Millipore is a registered trademark of Merck KGaA, Darmstadt, Germany
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service