Skip to Content
Merck
All Photos(1)

Key Documents

Z709298

ART® self-sealing barrier pipette tips

ART 500, volume range 50-500 μL, sterile

Synonym(s):

pipet tips, pipette tips

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41121601
NACRES:
NB.01
volume:
50—500 μL
sterility:
sterile
feature:
DNase free
RNase free
barrier
packaging:
pack of 800 ea (Racked)
manufacturer/tradename:
Molecular BioProducts 2275
Pricing and availability is not currently available.

material

colorless

sterility

sterile

feature

DNase free
RNase free
barrier

packaging

pack of 800 ea (Racked)

manufacturer/tradename

Molecular BioProducts 2275

technique(s)

PCR: suitable

maximum volume

500 μL

volume range

50-500 μL

volume

50—500 μL

compatibility

for use with

  • Eppendorf 4810 50-250 μl
  • Biohit Electronic 50-500 μl
  • Transferpette 25-250 μl

Looking for similar products? Visit Product Comparison Guide

General description

SoftFit design makes loading and ejecting easy

Packaging

960EA = 10 trays of 96 tips (1 pack)
4800EA = 5 packs of 960 tips (50 trays of 96 tips)

800EA = 8 trays of 100 tips (1pack)
3200EA = 4 packs of 800EA

Other Notes

Browse all pipette tips

Legal Information

ART is a registered trademark of Molecular BioProducts, Inc.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service