Skip to Content
Merck
All Photos(1)

Key Documents

Z614041

Vivaspin 500 centrifugal concentrators

MWCO 30 kDa

Synonym(s):

500 ml Vivaspin Centrifuges, Centrifuges, Vivaspin 500

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41104916
NACRES:
NB.24
Pricing and availability is not currently available.

material

polycarbonate housing
polyethersulfone membrane

description

PES membrane

sterility

non-sterile

feature

holdup volume 5 μL

packaging

package of 25 ea

manufacturer/tradename

Sartorius VS0121

parameter

15000 × g max. centrifugal force

dead stop volume

5 μL

membrane area

0.5 cm2

tube size

~2.2 mL

working volume

500 μL

pore size

30 kDa MWCO

Looking for similar products? Visit Product Comparison Guide

General description

  • Vivaspin centrifugal concentrators offer the optimal solution to any concentration or buffer exchange application with their broad range of volume capacities and MWCOs.
  • Highest flow rates and recoveries are achieved due to their particular vertical membrane design.
  • Incomparably safe sample handling and convenience is assured by the patented dead-stop technology allowing direct recovery of the concentrate without further re-spin.

Note: Vivaspin 500 μL centrifugal filter units offer a simple one step procedure for sample preparation. They can effectively be used in either swing out or fixed angle rotors.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service