Z331767
BRAND® pipette
withdraw volume 3.5 mL (including bulb), graduations, 3 mL (subdivision 0.5 mL), drop quantity 22 per mL
Synonym(s):
BRAND Pipette, Pipetting System
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
material
PE-LD
sterility
non-sterile
feature
graduations 3 mL (subdivision 0.5 mL)
packaging
pack of 500
pkg of 500 ea
manufacturer/tradename
BRAND 747760
withdraw volume measuring range
3.5 mL (including bulb)
L
154 mm
tip O.D.
3.2 mm
Looking for similar products? Visit Product Comparison Guide
General description
Disposable. Very good reproducibility of the number of drops per mL. Ideal for aliquots. Can be deep-frozen in filled state or changed into a closed vessel by heat-sealing the tip. The integrated pipetting bulb depresses easily, minimize fatigue from frequent pipetting.
Please note: can be sterilized by gas or γ-irradiation
Please note: can be sterilized by gas or γ-irradiation
Other Notes
BR747760 (India)
Legal Information
BRAND is a registered trademark of BRAND GMBH + CO KG
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service