Skip to Content
Merck
All Photos(1)

Key Documents

Y1271

Sigma-Aldrich

YM-202074 sesquifumarate salt hydrate

≥98% (HPLC)

Synonym(s):

N-cyclohexyl-6-{[(2-methoxyethyl)(methyl)amino]methyl}-N-methylthiazolo[3,2-a]benzimidazole-2-carboxamide, YM 202074, YM202074

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C22H30N4O2S · 1.5C4H4O4 · xH2O
CAS Number:
Molecular Weight:
588.67 (anhydrous basis)
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to off-white

solubility

DMSO: >20 mg/mL
H2O: >5 mg/mL

storage temp.

room temp

SMILES string

O.OC(=O)\C=C\C(O)=O.OC(=O)\C=C\C(O)=O.COCCN(C)Cc1ccc2nc3sc(cn3c2c1)C(=O)N(C)C4CCCCC4.COCCN(C)Cc5ccc6nc7sc(cn7c6c5)C(=O)N(C)C8CCCCC8.COCCN(C)Cc9ccc%10nc%11sc(cn%11c%10c9)C(=O)N(C)C%12CCCCC%12

InChI

1S/3C22H30N4O2S.2C4H4O4.H2O/c3*1-24(11-12-28-3)14-16-9-10-18-19(13-16)26-15-20(29-22(26)23-18)21(27)25(2)17-7-5-4-6-8-17;2*5-3(6)1-2-4(7)8;/h3*9-10,13,15,17H,4-8,11-12,14H2,1-3H3;2*1-2H,(H,5,6)(H,7,8);1H2/b;;;2*2-1+;

InChI key

MCGFIZFYSARKON-RAMYONIISA-N

Biochem/physiol Actions

YM-202074 is a potent and selective allosteric metabotropic glutamate receptor type 1 (mGluR1) antagonist. It bound an allosteric site of rat mGluR1 with a Ki value of 4.8 nM. It also inhibited the mGluR1-mediated inositol phosphates production in rat cerebellar granule cells with an IC50 value of 8.6 nM, while showing selectivity over mGluR(2-7).

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 3 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Despoina Goniotaki et al.
PLoS pathogens, 13(11), e1006733-e1006733 (2017-11-28)
Prion infections cause inexorable, progressive neurological dysfunction and neurodegeneration. Expression of the cellular prion protein PrPC is required for toxicity, suggesting the existence of deleterious PrPC-dependent signaling cascades. Because group-I metabotropic glutamate receptors (mGluR1 and mGluR5) can form complexes with

Articles

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Sigma-Aldrich offers many products related to G-protein family glutamate receptors for your research needs.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service