Skip to Content
Merck
All Photos(2)

Key Documents

WH0057348M4

Sigma-Aldrich

Monoclonal Anti-TTYH1 antibody produced in mouse

clone 4A9, purified immunoglobulin, buffered aqueous solution

Synonym(s):

Anti-tweety homolog 1 (Drosophila)

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

mouse

Quality Level

conjugate

unconjugated

antibody form

purified immunoglobulin

antibody product type

primary antibodies

clone

4A9, monoclonal

form

buffered aqueous solution

species reactivity

human

technique(s)

indirect ELISA: suitable
western blot: 1-5 μg/mL

isotype

IgG2aκ

GenBank accession no.

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... TTYH1(57348)

General description

This gene encodes a member of the tweety family of proteins. Members of this family function as chloride anion channels. The encoded protein functions as a calcium(2+)-independent, volume-sensitive large conductance chloride(-) channel. Two transcript variants encoding distinct isoforms have been identified for this gene. (provided by RefSeq)

Immunogen

TTYH1 (NP_065710, 261 a.a. ~ 360 a.a) partial recombinant protein with GST tag. MW of the GST tag alone is 26 KDa.

Sequence
MGLEAATAVGLSDFCSNPDPYVLNLTQEETGLSSDILSYYLLCNRAVSNPFQQRLTLSQRALANIHSQLLGLEREAVPQFPSAQKPLLSLEETLNVTEGN

Biochem/physiol Actions

The gene TTYH1 (tweety homolog 1) encodes a protein that functions as a chloride channel. It may be involved in early embryonic development. It is suggested to function in cell process formation and cell adhesion. Its expression in brain is found to be increased during epileptogenesis and epilepsy, indicating its involvement in brain pathology.

Physical form

Solution in phosphate buffered saline, pH 7.4

Legal Information

GenBank is a registered trademark of United States Department of Health and Human Services

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

nwg

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marzena Stefaniuk et al.
Journal of neurochemistry, 115(5), 1183-1194 (2010-09-30)
We have previously shown that Ttyh1 mRNA is expressed in neurons and its expression is up-regulated in the brain during epileptogenesis and epilepsy. In this study, we aimed to elucidate the role of Ttyh1 in neurons. We found widespread expression
Expression of Ttyh1, a member of the Tweety family in neurons in vitro and in vivo and its potential role in brain pathology.
Stefaniuk M
Journal of Neurochemistry, 115, 1183-1194 (2010)
Malgorzata Gorniak-Walas et al.
Neurochemical research, 46(9), 2463-2472 (2021-06-27)
Tweety-homolog 1 protein (Ttyh1) is abundantly expressed in neurons in the healthy brain, and its expression is induced under pathological conditions. In hippocampal neurons in vitro, Ttyh1 was implicated in the regulation of primary neuron morphology. However, the mechanisms that
Elzbieta Wiernasz et al.
Neurochemical research, 39(12), 2516-2526 (2014-10-16)
In a previous study, we showed that Ttyh1 protein is expressed in neurons in vitro and in vivo in the form of punctuate structures, which are localized to neuropil and neuronal somata. Herein, we provide the first description of Ttyh1
Juwan Kim et al.
EMBO reports, 19(11) (2018-09-05)
Despite growing evidence linking Drosophila melanogaster tweety-homologue 1 (Ttyh1) to normal mammalian brain development and cell proliferation, its exact role has not yet been determined. Here, we show that Ttyh1 is required for the maintenance of neural stem cell (NSC)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service