SYNS371208
Synthware™ Glass syringe
borosilicate glass 3.3, capacity 100 mL
Synonym(s):
Synthware S371208, glass sample syringe
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Recommended Products
material
borosilicate glass 3.3
packaging
pack of 1 ea
manufacturer/tradename
Kemtech S371208
capacity
100 mL
Looking for similar products? Visit Product Comparison Guide
General description
Glass Syringe (For Lab Use Only), Capacity: 100mL
Application
kemetch; synthware; SYRINGE
Features and Benefits
Organic Chemistry
Legal Information
Synthware is a trademark of Kemtech America Inc.
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Sorry, we don't have COAs for this product available online at this time.
If you need assistance, please contact Customer Support.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service