Skip to Content
Merck
All Photos(1)

Key Documents

SML3471

Sigma-Aldrich

MSC-5350

≥98% (HPLC)

Synonym(s):

5-[2-[5-Chloro-2-[[(5-methoxy-8-quinolinyl)sulfonyl]amino]phenyl]ethynyl]-4-methoxy-N,N-dimethyl-2-pyridinecarboxamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C27H23ClN4O5S
CAS Number:
Molecular Weight:
551.01
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear (Warmed)

storage temp.

2-8°C

SMILES string

O=C(N(C)C)C1=NC=C(C(OC)=C1)C#CC2=CC(Cl)=CC=C2NS(=O)(C3=CC=C(C4=CC=CN=C43)OC)=O

Biochem/physiol Actions

MSC-5350 is a cell penetrant negative control of MCT4 (monocarboxylate transporter 4 ) probe MSC-4381.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Discovery of 5-{2-[5-Chloro-2-(5-ethoxyquinoline-8-sulfonamido)phenyl]ethynyl}-4-methoxypyridine-2-carboxylic Acid, a Highly Selective in Vivo Useable Chemical Probe to Dissect MCT4 Biology
Journal of Medicinal Chemistry, 64 (2021)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service