Skip to Content
Merck
All Photos(1)

Key Documents

SML2450

Sigma-Aldrich

BIIE 0246 hydrochloride

≥98% (HPLC)

Synonym(s):

AR-H 053591 hydrochloride, BIIE-0246 hydrochloride, N-[(1S)-4-[(Aminoiminomethyl)amino]-1-[[[2-(3,5-dioxo-1,2-diphenyl-1,2,4-triazolidin-4-yl)ethyl]amino]carbonyl]butyl]-1-[2-[4-(6,11-dihydro-6-oxo-5H-dibenz[b,e]azepin-11-yl)-1-piperazinyl]-2-oxoethyl]cyclopentaneacetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C49H57N11O6 · xHCl
CAS Number:
Molecular Weight:
896.05 (free base basis)
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

storage condition

desiccated

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

2-8°C

SMILES string

[n]2([n]([c]([n]([c]2=O)CCNC(=O)[C@@H](NC(=O)CC8(CCCC8)CC(=O)N4CCN(CC4)C5c6c(cccc6)NC(=O)c7c5cccc7)CCCNC(=N)N)=O)c3ccccc3)c1ccccc1

InChI

1S/C49H57N11O6/c50-46(51)53-25-13-22-40(45(64)52-26-27-58-47(65)59(34-14-3-1-4-15-34)60(48(58)66)35-16-5-2-6-17-35)54-41(61)32-49(23-11-12-24-49)33-42(62)56-28-30-57(31-29-56)43-36-18-7-8-19-37(36)44(63)55-39-21-10-9-20-38(39)43/h1-10,14-21,40,43H,11-13,22-33H2,(H,52,64)(H,54,61)(H,55,63)(H4,50,51,53)/t40-,43?/m0/s1

InChI key

RSJAXPUYVJKAAA-JPGJPTAESA-N

Biochem/physiol Actions

BIIE0246 is a high-affinity, competitive non-peptide antagonist for the neuropeptide Y (NPY) Y2 receptor (Ki = 8-15 nM). BIIE0246 potently blocks NPY effects in rat hippocampus.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Bouchaïb El Bahh et al.
British journal of pharmacology, 136(4), 502-509 (2002-06-11)
Neuropeptide Y (NPY) has been shown to suppress synaptic excitation in rat hippocampus by a presynaptic action. The Y(2) (Y(2)R) and the Y(5) (Y(5)R) receptors have both been implicated in this action. We used the non-peptide, Y(2)R-selective antagonist, BIIE0246, to
C Lu et al.
Oncogene, 29(41), 5630-5642 (2010-08-03)
Neuroblastomas are pediatric tumors that develop from sympathetic precursors and express neuronal proteins, such as neuropeptide Y (NPY). NPY is a sympathetic neurotransmitter acting via multiple receptors (Y1-Y5R). Both NPY and Y2Rs are commonly expressed in neuroblastoma cell lines and
Y Dumont et al.
British journal of pharmacology, 129(6), 1075-1088 (2000-03-22)
1. BIIE0246, a newly synthesized non-peptide neuropeptide Y (NPY) Y(2) receptor antagonist, was able to compete with high affinity (8 to 15 nM) for specific [(125)I]PYY(3 - 36) binding sites in HEK293 cells transfected with the rat Y(2) receptor cDNA

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service