Skip to Content
Merck
All Photos(1)

Documents

SML2143

Sigma-Aldrich

Ailanthone

≥98% (HPLC)

Synonym(s):

Δ13-Dehydrochaparrinone, (1β,11β,12α)-11,20-Epoxy-1,11,12-trihydroxypicrasa-3,13(21)-diene-2,16-dione, 13,21-Didehydrochaparrinone

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C20H24O7
CAS Number:
Molecular Weight:
376.40
EC Number:
UNSPSC Code:
12352200
NACRES:
NA.77

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 2 mg/mL, clear

storage temp.

−20°C

SMILES string

O[C@@H]1C([C@]([C@@]2([C@]3([H])[C@@]1(O)OC2)[C@@](O4)([H])C[C@@]([C@]3(C)[C@@H]5O)([H])C(C)=CC5=O)([H])CC4=O)=C

InChI

1S/C20H24O7/c1-8-4-12(21)16(24)18(3)10(8)5-13-19-7-26-20(25,17(18)19)15(23)9(2)11(19)6-14(22)27-13/h4,10-11,13,15-17,23-25H,2,5-7H2,1,3H3/t10-,11-,13+,15+,16+,17+,18+,19+,20-/m0/s1

InChI key

WBBVXGHSWZIJST-RLQYZCPESA-N

Biochem/physiol Actions

Ailanthone, natural compound isolated from Ailanthus altissima (Simaroubaceae), is a potent inhibitor of androgen receptor (AR) that blocks activities of full-length AR and AR splice variants. It overcomes MDV3100 (Enzalutamide)-resistance in castration-resistant prostate cancer cell lines. Ailanthone disrupts the interaction between AR and the chaperones HSP90, HSP70, and HSP40, which leads to AR ubiquitination and degradation.

Pictograms

Skull and crossbones

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 2 Oral

Storage Class Code

6.1A - Combustible acute toxic Cat. 1 and 2 / very toxic hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yundong He et al.
Nature communications, 7, 13122-13122 (2016-12-14)
Androgen receptor (AR) antagonist MDV3100 is the first therapeutic approach in treating castration-resistant prostate cancer (CRPC), but tumours frequently become drug resistant via multiple mechanisms including AR amplification and mutation. Here we identify the small molecule Ailanthone (AIL) as a
Ailanthone: a new potential drug for castration-resistant prostate cancer.
Shihong Peng et al.
Chinese journal of cancer, 36(1), 25-25 (2017-03-05)
Zhenjian Zhuo et al.
Scientific reports, 5, 16185-16185 (2015-11-04)
While searching for natural anti-hepatocellular carcinoma (HCC) components in Ailanthus altissima, we discovered that ailanthone had potent antineoplastic activity against HCC. However, the molecular mechanisms underlying the antitumor effect of ailanthone on HCC have not been examined. In this study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service