Skip to Content
Merck
All Photos(1)

Key Documents

SML1890

Sigma-Aldrich

MK204

≥98% (HPLC)

Synonym(s):

2-[5-Chloro-2-[[[(2,3,4,5,6-pentabromophenyl)methyl]amino]carbonyl]phenoxy]-acetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C16H9Br5ClNO4
CAS Number:
Molecular Weight:
714.22
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 3 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

O=C(C1=C(OCC(O)=O)C=C(C=C1)Cl)NCC(C(Br)=C(C(Br)=C2Br)Br)=C2Br

InChI

1S/C16H9Br5ClNO4/c17-11-8(12(18)14(20)15(21)13(11)19)4-23-16(26)7-2-1-6(22)3-9(7)27-5-10(24)25/h1-3H,4-5H2,(H,23,26)(H,24,25)

InChI key

QYSFXUVFRUYJCZ-UHFFFAOYSA-N

Biochem/physiol Actions

MK204 is a potent and selective inhibitor of aldo-keto reductase family member 1B10 (AKR1B10). AKR1B10 is induced in several cancer types, in particular, hepatocellular carcinoma, and may be involved with drug resistance. MK204 appears to target a distinct AKR1B10 inner specificity pocket not present in the closely related aldose reductase. MK204 had an IC50 value of 80 nM compared to an IC50 value of 21.7 μM for aldose reductase.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Alexandra Cousido-Siah et al.
ACS chemical biology, 11(10), 2693-2705 (2016-10-22)
Human enzyme aldo-keto reductase family member 1B10 (AKR1B10) has evolved as a tumor marker and promising antineoplastic target. It shares high structural similarity with the diabetes target enzyme aldose reductase (AR). Starting from the potent AR inhibitor IDD388, we have

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service