Skip to Content
Merck
All Photos(1)

Key Documents

SML1058

Sigma-Aldrich

PBIT

≥98% (HPLC)

Synonym(s):

2-(4-Methylphenyl)-1,2-benzisothiazol-3(2H)-one; 2-(4-Methylphenyl)-2,3-dihydrobenzisothiazol-3-one, 2-p-Tolyl-1,2-benzisothiazolin-3-one

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H11NOS
CAS Number:
Molecular Weight:
241.31
UNSPSC Code:
12352200
NACRES:
NA.77

Quality Level

Assay

≥98% (HPLC)

form

powder

color

white to beige

solubility

DMSO: 10 mg/mL, clear (warmed)

storage temp.

2-8°C

SMILES string

[s]1[n]([c](c3c1cccc3)=O)c2ccc(cc2)C

InChI

1S/C14H11NOS/c1-10-6-8-11(9-7-10)15-14(16)12-4-2-3-5-13(12)17-15/h2-9H,1H3

InChI key

KRXMYBAZKJBJAB-UHFFFAOYSA-N

Related Categories

Biochem/physiol Actions

S,S′-1,4-phenylenebis(1,2-ethanediyl)bis-isothiourea (PBIT) is an iNOS (inducible nitric oxide synthase) inhibito. It is capable of inhibiting the multiplication of breast cancer cell lines expressing high levels of JARIDB1.
PBIT is a potent inhibitor of JARID1B (KDM5B) histone lysine demethylase that inhibits removal of H3K4me3 in UACC-812 cells. PBIT inhibits proliferation of cells overexpressing JARID1B. PBIT is selective for JARID1 enzymes and does not inhibit demethylases UTX or JMJD3.
PBIT is a potent inhibitor of JARID1B (KDM5B).

Features and Benefits

This compound is a featured product for Gene Regulation research. Click here to discover more featured Gene Regulation products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Slide 1 of 1

1 of 1

The Inhibitor Index (2017)
Melanomas: New Insights for the Healthcare Professional (2013)
Tharcisio Citrangulo Tortelli et al.
Aging, 13(18), 21914-21940 (2021-09-17)
Metformin has been tested as an anti-cancer therapy with potential to improve conventional chemotherapy. However, in some cases, metformin fails to sensitize tumors to chemotherapy. Here we test if the presence of P53 could predict the activity of metformin as

Articles

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

We offer a variety of small molecule research tools, such as transcription factor modulators, inhibitors of chromatin modifying enzymes, and agonists/antagonists for target identification and validation in gene regulation research; a selection of these research tools is shown below.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service