Skip to Content
Merck
All Photos(1)

Key Documents

SML0211

Sigma-Aldrich

iCRT3

Synonym(s):

2-[[[2-(4-ethylphenyl)-5-methyl-4-oxazolyl]methyl]thio]-N-(2-phenylethyl)acetamide

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C23H26N2O2S
CAS Number:
Molecular Weight:
394.53
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98% (HPLC)

Quality Level

form

powder

storage condition

desiccated

color

white to tan

solubility

DMSO: ≥5 mg/mL

storage temp.

2-8°C

SMILES string

CCc1ccc(cc1)-c2nc(CSCC(=O)NCCc3ccccc3)c(C)o2

InChI

1S/C23H26N2O2S/c1-3-18-9-11-20(12-10-18)23-25-21(17(2)27-23)15-28-16-22(26)24-14-13-19-7-5-4-6-8-19/h4-12H,3,13-16H2,1-2H3,(H,24,26)

InChI key

QTDYVSIBWGVBKU-UHFFFAOYSA-N

General description

iCRT3 is a small cell-permeable oxazole compound. It blocks cytokine secretion in lipopolysaccharide (LPS)-stimulated macrophages.

Application

iCRT3 has been used to inhibit the interactions between β-catenin and transcription factor (TCF).
iCRT3 has been used:
  • for β- catenin inhibition in dual luciferase assay
  • to inhibit β-catenin /TCF interactions and their transcriptional activity
  • as wingless/tailess (wnt) antagonist, to determine Wnt/β-catenin signaling is essential for PROX1-mediated regulation of forkhead box protein C2 (FOXC2)(3)catenin inhibition in dual luciferase assay
  • to inhibit β- catenin /TCF interactions and their transcriptional activity
  • as wingless/tailess (wnt) antagonist, to determine Wnt/β-catenin signaling is essential for PROX1-mediated regulation of forkhead box protein C2 (FOXC2)

Biochem/physiol Actions

The key mediator of Wnt signaling is the transcriptional co-activator b-catenin. In the cytoplasm, b-catenin is tightly bound to a complex that includes Axin and GSK-3b. Stimulation causes b-catenin stabilization, translocation to the nucleus and association with TCF4 to initiate transcription of responsive genes, referred to as Catenin Responsive Transcription (CRT). Virtually all Wnt-associated cancers are the result of misregulated CRT. Three inhibitors of CRT (iCRT) were identified in a screen that employed RNAi based knockdown of Axin, which stimulates CRT without affecting upstream mechanisms such as GSK activity, transduction by disheveled / frizzled, etc. These compounds are potent inhibitors of CRT reporter genes, as well as endogenous gene targets. The compounds also disrupt b-catenin-TCF4 interaction in a dose dependent manner, and cause G0/G1 arrest in colon tumor lines.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Mitigation of sepsis-induced inflammatory responses and organ injury through targeting Wntbeta-catenin signaling
Sharma A, et al.
Scientific reports, 7(1), 9235-9235 (2017)
Increasing beta-catenin/Wnt3A activity levels drive mechanical strain-induced cell cycle progression through mitosis
Benham-Pyle BW, et al.
eLife, 5, e19799-e19799 (2016)
Archna Sharma et al.
Scientific reports, 7(1), 9235-9235 (2017-08-25)
The Wnt/β-catenin pathway has been involved in regulating inflammation in various infectious and inflammatory diseases. Sepsis is a life-threatening condition caused by dysregulated inflammatory response to infection with no effective therapy available. Recently elevated Wnt/β-catenin signaling has been detected in
UVB represses melanocyte cell migration and acts through beta-catenin
Bertrand JU, et al.
Experimental Dermatology, 26(10), 875-882 (2017)
Complementary Wnt Sources Regulate Lymphatic Vascular Development via PROX1-Dependent Wntbeta-Catenin Signaling
Cha B, et al.
Testing, 25(3), 571-584 (2018)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service