Skip to Content
Merck
All Photos(1)

Key Documents

SMB01007

Sigma-Aldrich

Protoplumericin A

≥90% (LC/MS-ELSD)

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C36H42O19
CAS Number:
Molecular Weight:
778.71
MDL number:
UNSPSC Code:
12352205
NACRES:
NA.25

biological source

plant

Assay

≥90% (LC/MS-ELSD)

form

solid

mol wt

778.71

solubility

water: soluble

application(s)

metabolomics
vitamins, nutraceuticals, and natural products

storage temp.

−20°C

SMILES string

O1C(C(C(C(C1CO)O)O)O)O[C@@H]2OC=C([C@@H]3[C@H]2[C@@]4(OC(=O)C(=C4)[C@@H](OC(=O)\C=C\c5ccc(cc5)OC6OC(C(C(C6O)O)O)CO)C)C=C3)C(=O)OC

InChI

1S/C36H42O19/c1-15(50-23(39)8-5-16-3-6-17(7-4-16)51-34-29(44)27(42)25(40)21(12-37)52-34)19-11-36(55-32(19)47)10-9-18-20(31(46)48-2)14-49-33(24(18)36)54-35-30(45)28(43)26(41)22(13-38)53-35/h3-11,14-15,18,21-22,24-30,33-35,37-38,40-45H,12-13H2,1-2H3/b8-5+/t15-,18+,21?,22?,24+,25?,26?,27?,28?,29?,30?,33-,34?,35?,36+/m0/s1

InChI key

AFYIWKNGSIYXCQ-PYQCQGLYSA-N

General description

Protoplumericin A, an iridoid terpenoid, is commonly derived from Plumeria Sp. and Allamanda Sp. plants. This natural product, sourced from plants, has demonstrated anti-diabetic, anti-inflammatory, anticancer, and antioxidant properties. Additionally, it exhibits antibacterial activity against Gram-positive bacteria and fungi.

Application

It is a natural product derived from plant source that finds application in compound screening libraries, metabolomics, phytochemical, and biochemical research.

Biochem/physiol Actions

Protoplumericin A exhibits strong activity against S. aureus.

Features and Benefits

  • High quality compound suitable for multiple research applications
  • Compatible with HPLC and mass spectrometry techniques

Other Notes

For additional information on our range of Biochemicals, please complete this form.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Iridoids with antimicrobial activity from Plumeria alba l.
Afifi M S, et al.
Bulletin. American Pharmaceutical Association. Committee on National Formulary, 29, 215-223 null

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service