SAB4200403
Anti-EXOSC8 (N-terminal) antibody produced in rabbit
~1.0 mg/mL, affinity isolated antibody
Synonym(s):
Anit-CIP3, Anit-Exosome component 8,p9, Anti-EAP2, Anti-OIP2, Anti-Opa interacting protein 2, Anti-RP11-421P11.3, Anti-RRP43, Anti-Ribosomal RNA-processing protein 43, Anti-Rrp43p, AntiCBP-interacting protein 3
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
mol wt
antigen ~34 kDa
species reactivity
mouse, human, rat
concentration
~1.0 mg/mL
technique(s)
immunoprecipitation (IP): 1-2 μg using human HEK-293T cell lysate
western blot: 1-2 μg/mL using whole extracts of mouse A20 and rat PC12 cells
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
bovine ... EXOSC8(532080)
dog ... EXOSC8(477300)
human ... EXOSC8(11340)
mouse ... Exosc8(69639)
General description
Specificity
Immunogen
Application
Biochem/physiol Actions
Physical form
Storage and Stability
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Certificates of Analysis (COA)
Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service