Skip to Content
Merck
All Photos(1)

Key Documents

SAB2101135

Sigma-Aldrich

Anti-IFT172 antibody produced in rabbit

affinity isolated antibody

Synonym(s):

Anti-DKFZP434A163, Anti-Intraflagellar transport 172 homolog (Chlamydomonas), Anti-KIAA1179, Anti-Osm-1, Anti-SLB

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41
conjugate:
unconjugated
application:
WB
clone:
polyclonal
species reactivity:
dog, mouse, horse, bovine, human, rabbit, guinea pig, rat
citations:
technique(s):
western blot: suitable

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

mol wt

197 kDa

species reactivity

dog, mouse, horse, bovine, human, rabbit, guinea pig, rat

concentration

0.5 mg - 1 mg/mL

technique(s)

western blot: suitable

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... IFT172(26160)

Immunogen

Synthetic peptide directed towards the N terminal region of human IFT172

Biochem/physiol Actions

IFT172 is required for the maintenance and formation of cilia. IT plays an indirect role in hedgehog (Hh) signaling, cilia being required for all activity of the hedgehog pathway.

Sequence

Synthetic peptide located within the following region: VKILGKERYLVAHTSETLLLGDLNTNRLSEIAWQGSGGNEKYFFENENVC

Physical form

Purified antibody supplied in 1x PBS buffer with 0.09% (w/v) sodium azide and 2% sucrose.

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service