SAB1300765
Anti-PARP16 (C-term) antibody produced in rabbit
saturated ammonium sulfate (SAS) precipitated, buffered aqueous solution
Synonym(s):
Anti-EC 2.4.2.30, Anti-PARP-16, Anti-Poly [ADP-ribose] polymerase 16
About This Item
Recommended Products
biological source
rabbit
Quality Level
conjugate
unconjugated
antibody form
saturated ammonium sulfate (SAS) precipitated
antibody product type
primary antibodies
clone
polyclonal
form
buffered aqueous solution
species reactivity
mouse, human
technique(s)
indirect ELISA: 1:1000
western blot: 1:100-1:500
NCBI accession no.
UniProt accession no.
shipped in
dry ice
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... PARP16(214424)
General description
Immunogen
This antibody is generated from rabbits immunized with a KLH-conjugated synthetic peptide selected from the C-terminal region of mouse PARP16.
Physical form
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
10 - Combustible liquids
WGK
nwg
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service