S7826
SMER3
≥98% (HPLC)
Synonym(s):
9H-Indeno-[1,2-e]-[1,2,5]-oxadiazolo-[3,4-b]-pyrazin-9-one
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Empirical Formula (Hill Notation):
C11H4N4O2
CAS Number:
Molecular Weight:
224.18
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77
Recommended Products
Assay
≥98% (HPLC)
form
powder
color
yellow
solubility
DMSO: ≥30 mg/mL
storage temp.
2-8°C
SMILES string
O=C1c2ccccc2-c3nc4nonc4nc13
InChI
1S/C11H4N4O2/c16-9-6-4-2-1-3-5(6)7-8(9)13-11-10(12-7)14-17-15-11/h1-4H
InChI key
SFSSAKVWCKFRHE-UHFFFAOYSA-N
Application
SMER3 may be used in cell signaling studies.
Biochem/physiol Actions
Inhibition of SCFMet30 by SMER3 blocks the ubiquitination of Met4 and activates the Met4-containing transcription complex. The induction of MET-genes results in cell cycle arrest and reduced cell proliferation.1
Small molecule enhancer of rapamycin (SMER); specific inhibitor of Ubiquitin E3 ligase.
Storage Class Code
11 - Combustible Solids
WGK
WGK 1
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Mariam Aghajan et al.
Nature biotechnology, 28(7), 738-742 (2010-06-29)
The target of rapamycin (TOR) plays a central role in eukaryotic cell growth control. With prevalent hyperactivation of the mammalian TOR (mTOR) pathway in human cancers, strategies to enhance TOR pathway inhibition are needed. We used a yeast-based screen to
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service