Skip to Content
Merck
All Photos(1)

Key Documents

RAB1480

Sigma-Aldrich

Human GDF-11 ELISA

for serum, plasma and cell culture supernatants

Synonym(s):

BMP11, Bone morphogenetic protein 11

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41116158
NACRES:
NA.84

species reactivity

human

packaging

kit of 96 wells (12 strips x 8 wells)

technique(s)

ELISA: suitable

input

sample type plasma
sample type cell culture supernatant(s)
sample type serum

assay range

inter-assay cv: <12%
intra-assay cv: <10%
sensitivity: 0.16 ng/mL
standard curve range: 0.164-40 ng/mL

detection method

colorimetric

shipped in

wet ice

storage temp.

−20°C

Gene Information

human ... GDF11(10220)

General description

Growth differentiation factor 11 (GDF11), also known as bone morphogenetic protein 11 (BMP11) is a member of the BMP/transforming growth factor β (TGF-β) superfamily. GDF11 consists of a signaling sequence, an RXXR proteolytic processing site, and a conserved cysteine-containing a C-terminal end. It is expressed in several tissues associated with the kidney, pancreas, retina, and nervous system. GDF11 plays an important role in embryonic development, brain function, and signal transduction. It is involved in tumor suppression of breast and liver cancer cells.

Application

For research use only. Not for use in diagnostic procedures.
Please refer to the attached Protocolfor details.

Other Notes

A sample Certificate of Analysis is available for this product. Please type the word sample in the text box provided for lot number.

Pictograms

Corrosion

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Met. Corr. 1

Storage Class Code

8A - Combustible corrosive hazardous materials

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Yong-Hui Zhang et al.
Experimental and therapeutic medicine, 15(4), 3495-3500 (2018-03-17)
Growth differentiation factor 11 (GDF11), also known as bone morphogenetic protein 11, a member of the transforming growth factor-β superfamily, has been reported to be involved in colorectal cancer. However, the roles of GDF11 in Chinese patients with liver cancer

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service