Skip to Content
Merck
All Photos(1)

Key Documents

PRS4691

Sigma-Aldrich

Anti-GDF6 antibody produced in rabbit

affinity isolated antibody, buffered aqueous solution

Synonym(s):

Anti-CDMP2, Anti-GDF16, Anti-Growth differentiation factor 6

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

form

buffered aqueous solution

species reactivity

mouse, human, rat

technique(s)

indirect ELISA: suitable
western blot: suitable

UniProt accession no.

shipped in

dry ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... GDF6(392255)

General description

Growth differentiation factor 6 (GDF6) is a secreted signalling molecule, that belongs to the family of bone morphogenetic protein (BMP). Gdf6 is expressed in embryonic tissues that is concerned with patterning of skeletal and soft tissue. In human chromosome, the gene GDF6 is localized on 8q22.1.

Immunogen

a 17 amino acid peptide near the carboxy-terminus of the human GDF6.

Application

Anti-GDF6 antibody produced in rabbit has been used in western blotting.

Biochem/physiol Actions

Growth differentiation factor 6 (GDF6) plays a crucial role in the joint formation during skeletal development. Gdf6 is involved in the maintenance of articular cartilage of knee. Mutation of GDF6 leads to loss-of-function and causes Klippel-Feil syndrome. Microduplication of GDF6 leads to an autosomal-dominant rheumatic condition, Leri′s pleonosteosis (LP). Mutations in GDF6 causes Multiple synostoses syndrome subtype SYNS4. In SYNS4, there is fusion of joints leading to progressive conductive deafness. GDF6 plays a key role in ocular development and the mutation of GDF6 leads to microphthalmia and anophthalmia. Mutations in GDF6 also leads to Leber congenital amaurosis-17.

Linkage

The action of this antibody can be blocked using blocking peptide SBP4691.

Physical form

Solution in phosphate buffered saline containing 0.02% sodium azide

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

related product

Product No.
Description
Pricing

Storage Class Code

10 - Combustible liquids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

A new subtype of multiple synostoses syndrome is caused by a mutation in GDF6 that decreases its sensitivity to noggin and enhances its potency as a BMP signal
Wang J, et al.
Journal of Bone and Mineral Research, 31(4), 882-889 (2016)
Growth differentiation factor 6 derived from mesenchymal stem/stromal cells reduces age-related functional deterioration in multiple tissues
Hisamatsu D, et al.
Aging (Albany. NY.), 8(6), 1259-1259 (2016)
Eye and neural defects associated with loss of GDF6
Hanel ML and Hensey C
BMC Developmental Biology, 6(1), 43-43 (2006)
GDF6, a novel locus for a spectrum of ocular developmental anomalies
Asai-Coakwell M, et al.
American Journal of Human Genetics, 80(2), 306-315 (2007)
Daisuke Hisamatsu et al.
Aging, 8(6), 1259-1275 (2016-06-18)
The senescence-associated secretory phenotype (SASP) has attracted attention as a mechanism that connects cellular senescence to tissue dysfunction, and specific SASP factors have been identified as systemic pro-aging factors. However, little is known about the age-dependent changes in the secretory

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service