Skip to Content
Merck
All Photos(1)

Key Documents

P8035

Sigma-Aldrich

Poly(γ-ethyl-L-glutamate)

mol wt >100,000

Sign Into View Organizational & Contract Pricing


About This Item

CAS Number:
MDL number:
UNSPSC Code:
12352202
NACRES:
NA.26

form

powder or solid

mol wt

>100,000

technique(s)

bioconjugation: suitable

color

white to off-white

storage temp.

−20°C

Analysis Note

Molecular weight based on viscosity

Other Notes

For additional technical information on polyamino acids please visit the Polyamino acid FAQ resource.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Protonation of peptides. I. The behavior of a model diamide and of poly-gamma-ethyl-L-glutamate in strong acid-water mixtures.
J Steigman et al.
Journal of the American Chemical Society, 91(7), 1822-1829 (1969-03-26)
S A Mitchell et al.
The Journal of chemical physics, 122(11), 114707-114707 (2005-04-20)
Quantitative measurements of second-harmonic generation optical activity (SHG-OA) have been performed for alpha-helical polypeptides poly-(gamma-benzyl-L-glutamate) and poly-(gamma-ethyl-L-glutamate) adsorbed at the airwater interface, with the fundamental frequency variant Planck's over 2piomega = 2.96 eV (lambda = 417 nm). The chiral component
Christina M Thiele
The Journal of organic chemistry, 69(22), 7403-7413 (2004-10-23)
The concept of using residual dipolar couplings (RDCs) for the structure determination of organic molecules is applied to the simultaneous assignment of all diastereotopic protons in strychnine. To use this important NMR parameter the molecule has to be aligned in

Articles

Humankind has utilized protein materials throughout its existence, starting with the use of materials such as wool and silk for warmth and protection from the elements and continuing with the use of recombinant DNA techniques to synthesize proteins with unique and useful properties.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service