Skip to Content
Merck
All Photos(3)

Key Documents

P0778

Sigma-Aldrich

Pindolol

≥98% (TLC), powder

Synonym(s):

1-(1H-Indol-4-yloxy)-3-(isopropylamino)-2-propanol, 1-(1H-Indol-4-yloxy)-3-[(1-methylethyl)amino]-2-propanol

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C14H20N2O2
CAS Number:
Molecular Weight:
248.32
EC Number:
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Quality Level

Assay

≥98% (TLC)

form

powder

color

white to off-white

mp

167-171 °C (lit.)

solubility

0.1 M NaOH: 0.2 mg/mL
45% (w/v) aq 2-hydroxypropyl-β-cyclodextrin: 0.2 mg/mL
0.1 M HCl: 20 mg/mL
acetic acid: 50 mg/mL
H2O: insoluble

originator

Novartis

storage temp.

2-8°C

SMILES string

CC(C)NCC(O)COc1cccc2[nH]ccc12

InChI

1S/C14H20N2O2/c1-10(2)16-8-11(17)9-18-14-5-3-4-13-12(14)6-7-15-13/h3-7,10-11,15-17H,8-9H2,1-2H3

InChI key

JZQKKSLKJUAGIC-UHFFFAOYSA-N

Gene Information

Looking for similar products? Visit Product Comparison Guide

Application

Pindolol has been used in intestinal transport and metabolism assays and cassette mix preparation.

Biochem/physiol Actions

β1-adrenoceptor antagonist; putative 5-HT1A serotonin receptor agonist; vasodilator. Pindolol is a non-cardioselective beta-adrenergic blocking agent. It possesses intrinsic sympathomimetic activity.
β1-adrenoceptor antagonist; putative 5-HT1A serotonin receptor agonist; vasodilator.

Features and Benefits

Shelf-life of the powder is at least five years.
This compound is a featured product for ADME Tox and Neuroscience research. Discover more featured ADME Tox and Neuroscience products. Learn more about bioactive small molecules for other areas of research at sigma.com/discover-bsm.
This compound is featured on the β-Adrenoceptors page of the Handbook of Receptor Classification and Signal Transduction. To browse other handbook pages, click here.
This compound was developed by Novartis. To browse the list of other pharma-developed compounds and Approved Drugs/Drug Candidates, click here.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

The biotransformation of prasugrel, a new thienopyridine prodrug, by the human carboxylesterases 1 and 2
Williams E T, et al.
Drug Metabolism and Disposition (2008)
Melanin binding study of clinical drugs with cassette dosing and rapid equilibrium dialysis inserts
Pelkonen L, et al.
European Journal of Pharmaceutical Sciences, 162-168 (2017)
C M Hysek et al.
Emergency medicine journal : EMJ, 27(8), 586-589 (2010-04-10)
MDMA (3,4-methylenedioxymethamphetamine, 'Ecstasy') produces tachycardia and hypertension and is rarely associated with cardiovascular and cerebrovascular complications. In clinical practice, beta-blockers are often withheld in patients with stimulant intoxication because they may increase hypertension and coronary artery vasospasm due to loss
Pindolol: a review of its pharmacology, pharmacokinetics, clinical uses, and adverse effects
Golightly L K
Pharmacotherapy, 2(3), 134-147 (1982)
Maria J Portella et al.
The Journal of clinical psychiatry, 72(7), 962-969 (2010-11-03)
Since depression entails not only dramatic personal disruption but also a huge amount of medical and socioeconomic burden, slowness of antidepressant action and difficulties to attain remission are entangled issues to be solved. Given the controversial previous findings with enhancing

Articles

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

Discover Bioactive Small Molecules for ADME/Tox

See All

Chromatograms

application for HPLCapplication for HPLC

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service