Skip to Content
Merck
All Photos(1)

Key Documents

N7787

Sigma-Aldrich

NS3694

≥98%, powder

Synonym(s):

4-Chloro-2-[3-(3-trifluoromethyl-phenyl)-ureido]benzoic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C15H10ClF3N2O3
CAS Number:
Molecular Weight:
358.70
MDL number:
UNSPSC Code:
12352200
PubChem Substance ID:
NACRES:
NA.77

Assay

≥98%

form

powder

solubility

DMSO: soluble 22 mg/mL
H2O: insoluble <5 mg/mL

storage temp.

2-8°C

SMILES string

OC(=O)c1ccc(Cl)cc1NC(=O)Nc2cccc(c2)C(F)(F)F

InChI

1S/C15H10ClF3N2O3/c16-9-4-5-11(13(22)23)12(7-9)21-14(24)20-10-3-1-2-8(6-10)15(17,18)19/h1-7H,(H,22,23)(H2,20,21,24)

InChI key

GNCZTZCPXFDPLI-UHFFFAOYSA-N

Gene Information

human ... GRIK1(2897)

Application

NS3694 may be used as an apoptosis inhibitor to study the relationship between the cellular mechanisms controlled by amputation-induced myeloid lineage activity.

Biochem/physiol Actions

NS3694 can prevent the active apoptosome complex formation.
NS3694 is an Inhibitor of apoptosome formation.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Jovany J Franco et al.
Molecular biology of the cell, 30(16), 1890-1899 (2019-02-21)
Epithelial wound healing requires a complex orchestration of cellular rearrangements and movements to restore tissue architecture and function after injury. While it is well known that mechanical forces can affect tissue morphogenesis and patterning, how the biophysical cues generated after
Cindy X Kha et al.
Experimental eye research, 169, 38-47 (2018-01-23)
Vertebrate eye development is complex and requires early interactions between neuroectoderm and surface ectoderm during embryogenesis. In the African clawed frog, Xenopus laevis, individual eye tissues such as the retina and lens can undergo regeneration. However, it has been reported
Developmental dependence for functional eye regrowth in Xenopus laevis.
Cindy X Kha et al.
Neural regeneration research, 13(10), 1735-1737 (2018-08-24)
Ulrik Lademann et al.
Molecular and cellular biology, 23(21), 7829-7837 (2003-10-16)
The release of mitochondrial proapoptotic proteins into the cytosol is the key event in apoptosis signaling, leading to the activation of caspases. Once in the cytosol, cytochrome c triggers the formation of a caspase-activating protein complex called the apoptosome, whereas
Can Aztekin et al.
Development (Cambridge, England), 147(3) (2020-01-29)
Regeneration-competent vertebrates are considered to suppress inflammation faster than non-regenerating ones. Hence, understanding the cellular mechanisms affected by immune cells and inflammation can help develop strategies to promote tissue repair and regeneration. Here, we took advantage of naturally occurring tail

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service