Skip to Content
Merck
All Photos(2)

Key Documents

MAK008

Sigma-Aldrich

Hydroxyproline Assay Kit

sufficient for 100 colorimetric tests

Synonym(s):

Hydroxyproline Quantification Kit

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12161503
NACRES:
NA.84

usage

sufficient for 100 colorimetric tests

detection method

colorimetric

relevant disease(s)

dermatological diseases

shipped in

wet ice

storage temp.

2-8°C

General description

Hydroxyproline (4-hydroxyproline) is a non-proteinogenic amino acid formed by the post-translational hydroxylation of proline. Hydroxyproline is a major component of collagen, where it serves to stabilize the helical structure. Because hydroxyproline is largely restricted to collagen, the measurement of hydroxylproline levels can be used as an indicator of collagen content. Conditions that increase collagen turnover can elevate serum and urine hydroxyproline levels. Urine and serum hydroxyproline levels can be used as a marker for bone resorption.

Application

Hydroxyproline Assay Kit has been used to determine the concentration of hydroxyproline and collagen.

Suitability

Suitable for hydroxyproline detection in cell and tissue culture supernatants, urine, plasma, serum, and other biological samples.

Principle

In the Hydroxyproline Assay Kit, hydroxyproline concentration is determined by the reaction of oxidized hydroxyproline with 4-(Dimethylamino)benzaldehyde (DMAB), which results in a colorimetric (560 nm) product, proportional to the hydroxyproline present.

replaced by

Product No.
Description
Pricing

Signal Word

Danger

Hazard Classifications

Eye Dam. 1 - Flam. Liq. 2 - Met. Corr. 1 - Ox. Liq. 2 - Resp. Sens. 1 - Skin Corr. 1B - Skin Sens. 1 - STOT RE 2 - STOT SE 3

Target Organs

Central nervous system, Thyroid

Supplementary Hazards

Storage Class Code

5.1B - Oxidizing hazardous materials

Flash Point(F)

64.4 °F

Flash Point(C)

18 °C


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Lodish H, et al.
Molecular Cell Biology. 4th edition. (2000)
Urinary biochemical markers for bone resorption during the menstrual cycle.
Gorai I, et al.
Calcified Tissue International, 57(2), 100-104 (1995)
Chunming Jiang et al.
BioMed research international, 2015, 867140-867140 (2016-02-18)
Acute kidney injury (AKI) is associated with an increased risk of developing advanced chronic kidney disease (CKD). Yet, effective interventions to prevent this conversion are unavailable for clinical practice. In this study, we examined the beneficial effects of Tanshinone IIA
Krysta Powers et al.
The Journal of experimental biology, 220(Pt 17), 3110-3118 (2017-06-24)
In actively stretched skeletal muscle sarcomeres, titin-based force is enhanced, increasing the stiffness of active sarcomeres. Titin force enhancement in sarcomeres is vastly reduced in
Biochemical markers of bone turnover part I: biochemistry and variability.
Seibel M J.
The Clinical Biochemist. Reviews / Australian Association of Clinical Biochemists, 26(4), 97-97 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service