MAK007
Asparaginase Activity Assay Kit
sufficient for 100 colorimetric or fluorometric tests
Synonym(s):
Asparaginase Test Kit
Sign Into View Organizational & Contract Pricing
All Photos(4)
About This Item
UNSPSC Code:
12161503
NACRES:
NA.84
Recommended Products
usage
sufficient for 100 colorimetric or fluorometric tests
detection method
colorimetric
fluorometric
relevant disease(s)
cancer
storage temp.
−20°C
General description
Asparaginase is an enzyme that catalyzes the hydrolysis of asparagine to aspartate. While asparaginase is synthesized by plants, microorganisms, and some animals, it does not occur naturally in humans. While most cells have the ability to synthesize asparagine, many hematopoietic cells do not and depend on exogenous asparagine for protein synthesis. Treatment of certain hematopoietic malignancies such as acute lymphoblastic leukemia (ALL) with asparaginase results in depletion of blood levels of asparagine resulting in cell cycle arrest and apoptosis. Asparaginase is also used in the food industry to reduce the formation of acrylamide in starchy and fried foods.
Suitability
Suitable for measuring asparaginase activity in biological samples
Principle
The Asparaginase Activity Assay Kit provides a simple and direct procedure for measuring Asparaginse activity in a variety of biological samples. Asparaginase activity is determined by a coupled enzyme assay, which results in a colorimetric (570 nm)/fluorometric (λex = 535/λem = 587 nm) product, proportional to the aspartate generated. One milliunit (mU) of Asparaginase is defined as the amount of enzyme that catalyzes the formation of 1.0 mmole of aspartate per minute at 25 °C.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Resp. Sens. 1
Storage Class Code
10 - Combustible liquids
Flash Point(F)
188.6 °F - closed cup
Flash Point(C)
87 °C - closed cup
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Customers Also Viewed
Asparagine plays a critical role in regulating cellular adaptation to glutamine depletion.
.Zhang J, et al.
Molecular Cell, 56(2), 205-218 (2014)
Current applications and different approaches for microbial l-asparaginase production.
Cachumba J J M, et al.
Brazilian Journal of Microbiology, 47, 77-85 (2016)
Semi-permeable coatings fabricated from comb-polymers efficiently protect proteins in vivo.
Liu M, et al.
Nature Communications, 5, 5526-5526 (2014)
An inhibitor of human asparagine synthetase suppresses proliferation of an L-asparaginase-resistant leukemia cell line.
Gutierrez J A, et al.
Chemistry and Biology, 13(1), 1339-1347 (2006)
Yalin Zhang et al.
The American journal of Chinese medicine, 47(5), 1133-1147 (2019-07-18)
Hyperuricemia is a metabolic disease of the kidney that results in decreased uric acid excretion. Here, we aimed to investigate the effects of ginsenosides and anserine on hyperuricemia and the expression of aquaporin (AQP) 1-4, which are indicators of renal
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service