Skip to Content
Merck
All Photos(1)

Documents

M9197

Sigma-Aldrich

MLCK (1425-1776), active, GST tagged human

PRECISIO® Kinase, recombinant, expressed in baculovirus infected Sf9 cells, ≥70% (SDS-PAGE), buffered aqueous glycerol solution

Synonym(s):

DKFZp686I10125, FLJ12216, KRP, MLCK108, MLCK210, MSTP083, MYLK

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352200
NACRES:
NA.32

recombinant

expressed in baculovirus infected Sf9 cells

Quality Level

product line

PRECISIO® Kinase

Assay

≥70% (SDS-PAGE)

form

buffered aqueous glycerol solution

specific activity

170-230 nmol/min·mg

mol wt

~70 kDa

UniProt accession no.

shipped in

dry ice

storage temp.

−70°C

Gene Information

human ... MYLK(4638)

Biochem/physiol Actions

MLCK or myosin light chain kinase is a muscle member of the immunoglobulin gene superfamily and is a calcium/calmodulin dependent protein kinase. MLCK is a regulatory protein for smooth muscle contraction, which acts by phosphorylating 20-kDa myosin light chain (MLC20) to activate the myosin ATPase activity. The kinase activity for the phosphorylation is localized at the central part of MLCK, which is also furnished with actin-binding activity at its N terminal and myosin-binding activity at its C terminal. Phosphorylation of regulatory light chain of myosin plays an important role in controlling the morphological changes seen during cell division.

Physical form

Supplied in 50 mM Tris-HCl, pH 7.5, with 150 mM NaCl, 0.2 5mM DTT, 0.1 mM EGTA, 0.1 mM EDTA, 0.1 mM PMSF, and 25% glycerol.

Legal Information

PRECISIO is a registered trademark of Merck KGaA, Darmstadt, Germany

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

F Matsumura et al.
Cell structure and function, 26(6), 639-644 (2002-04-11)
Phosphorylation of regulatory light chain (RMLC) of myosin II at Ser19/Thr18 is likely to play important roles in controlling the morphological changes seen during cell division of cultured mammalian cells. Phosphorylation of RMLC regulates the activity of myosin II, an
Y Gao et al.
IUBMB life, 51(6), 337-344 (2002-01-05)
Myosin light chain kinase (MLCK) is a regulatory protein for smooth muscle contraction, which acts by phosphorylating 20-kDa myosin light chain (MLC20) to activate the myosin ATPase activity. Although this mode of action is well-established, there are numerous reports of
Tünde Juhász et al.
FASEB bioAdvances, 2(8), 489-505 (2020-08-22)
Calmodulin (CaM), the key calcium sensor of eukaryotic cells regulating a great number of target proteins, belongs to the most conserved proteins. We compared function and properties of CaMs from two evolutionarily distant species, the human (Homo sapiens) representing vertebrates
Kasturi Markandran et al.
International journal of molecular sciences, 23(1) (2022-01-12)
Heart failure (HF) as a result of myocardial infarction (MI) is a major cause of fatality worldwide. However, the cause of cardiac dysfunction succeeding MI has not been elucidated at a sarcomeric level. Thus, studying the alterations within the sarcomere

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service