HS14521B
Aluminium weighing boat
D × H 57 mm × 12 mm , meduim
All Photos(1)
About This Item
UNSPSC Code:
41121812
NACRES:
NB.35
Recommended Products
manufacturer/tradename
Heathrow Scientific HS14521B
D × H
57 mm × 12 mm , meduim
General description
designed for general-purpose weighing, dispensing, storage, food processing, evaporating, or drying.
Boats also make good dust covers for open vessels.
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
Boats also make good dust covers for open vessels.
- All weigh boats are FDA/food-grade compliant
- Disposable
- Aluminum, Weighing Boats
Sturdy yet lightweight boats feature a smooth, flat bottom for sample handling and a flanged, crimped wall for rigidity. Finger tab handle can be etched for identification prior to weighing.
3 sizes
Packs of 100
Choose from one of the most recent versions:
Certificates of Analysis (COA)