Skip to Content
Merck
All Photos(5)

Key Documents

HPA023589

Sigma-Aldrich

Anti-NAA15 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Gastric cancer antigen Ga19, Anti-N-terminal acetyltransferase, Anti-NARG1, Anti-NMDA receptor-regulated protein 1, Anti-Protein tubedown-1, Anti-Tbdn100

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:200-1:500

immunogen sequence

ALEHLCTYEKQICDKLAVEETKGELLLQLCRLEDAADVYRGLQERNPENWAYYKGLEKALKPANMLERLKIYEEAWTKYPRGLVPRRLPLNFLSGEKFKECLDKFLRMNFSKG

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... NARG1(80155)

General description

The gene NAA15 (N(α)-acetyltransferase 15, NatA auxiliary subunit), also referred to as NATH, has two mRNA species of 4.6 and 5.8kb size. They have the same open reading frame and encode a putative protein of 866 amino acids. The encoded protein contains four tetratricopeptide repeat (TPR) domains and may be a component of a multiprotein complex. It is homologous to yeast N-acetyltransferase (NAT)1 and to mouse NARG1 (mNAT1) proteins. It is expressed in small amounts in most adult tissues, including normal thyroid gland. It is found to be localized to the cytoplasm and has a molar mass of 105kDa.

Immunogen

NMDA receptor-regulated protein 1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

The expression of NAA15 (N(α)-acetyltransferase 15, NatA auxiliary subunit) is found to be higher in papillary thyroid carcinoma, suggesting its role in carcinogenesis. It is found to be associated with human arrest defective 1 (hARD1), an N-α-acetyltransferase, forming a complex that transfers acetyl groups to the N-termini of nascent polypeptides. This complex functions in cell survival and its knockdown or downregulation results in increased apoptosis.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST70962

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

NATH, a novel gene overexpressed in papillary thyroid carcinomas.
Fluge ?
Oncogene, 21, 5056-5068 (2002)
Induction of apoptosis in human cells by RNAi-mediated knockdown of hARD1 and NATH, components of the protein N-alpha-acetyltransferase complex.
Arnesen T
Oncogene, 25, 4350-4360 (2006)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service