Skip to Content
Merck
All Photos(1)

Key Documents

HPA020610

Sigma-Aldrich

Anti-SVEP1 antibody produced in rabbit

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution, Ab1

Synonym(s):

Anti-CCP module-containing protein 22, Anti-Polydom, Anti-SEL-OB, Anti-Selectin-like osteoblast-derived protein, Anti-Serologically defined breast cancer antigen NY-BR-38, Anti-Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.41

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

technique(s)

immunohistochemistry: 1:20- 1:50

immunogen sequence

NRLDYSYDDFLDTVQETATSIGNAKSSRIKRSAPLSDYKIKLIFNITASVPLPDERNDTLEWENQQRLLQTLETITNKLKRTLNKDPMYSFQLASEIL

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... SVEP1(79987)

General description

Sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1 (SVEP1) is expressed in osteogenic cells and has been shown to be expressed in skeletal tissues in vivo. The gene encoding it is localized on human chromosome 9q32.

Immunogen

Sushi, von Willebrand factor type A, EGF and pentraxin domain-containing protein 1 Precursor recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Sushi, von Willebrand factor type A, EGF and pentraxin domain containing 1 (SVEP1) regulates cell adhesion which depends on integrin α9β1. It might influence the proteins involved in the leukocyte adhesion pathway. SVEP1 might be useful as a marker to study the characteristics and functions of osteogenic cells.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST75351

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Taka-aki Nakada et al.
Critical care medicine, 43(1), 101-108 (2014-09-05)
Mortality from septic shock is highly heritable. The identification of causal genetic factors is insufficient. To discover key contributors, we first identified nonsynonymous single-nucleotide polymorphisms in conserved genomic regions that are predicted to have significant effects on protein function. We
I Shur et al.
Journal of cellular physiology, 206(2), 420-427 (2005-10-06)
We describe a novel human gene, named SEL-OB/SVEP1, expressed by skeletal tissues in vivo and by cultured osteogenic cells. The mRNA expression was analyzed on frozen tissues retrieved by laser-capture microscope dissection (LCM) and was detected in osteogenic tissues (periosteum
Michael J Winkler et al.
Basic research in cardiology, 115(6), 67-67 (2020-11-14)
A missense variant of the sushi, von Willebrand factor type A, EGF and pentraxin domain containing protein 1 (SVEP1) is genome-wide significantly associated with coronary artery disease. The mechanisms how SVEP1 impacts atherosclerosis are not known. We found endothelial cells
Ryoko Sato-Nishiuchi et al.
The Journal of biological chemistry, 287(30), 25615-25630 (2012-06-02)
A variety of proteins, including tenascin-C and osteopontin, have been identified as ligands for integrin α9β1. However, their affinities for integrin α9β1 are apparently much lower than those of other integrins (e.g. α3β1, α5β1, and α8β1) for their specific ligands
D Gilgès et al.
The Biochemical journal, 352 Pt 1, 49-59 (2000-11-04)
To identify extracellular proteins with epidermal growth factor (EGF) domains that are potentially involved in the control of haemopoiesis, we performed degenerate reverse-transcriptase-mediated PCR on the murine bone-marrow stromal cell line MS-5 and isolated a new partial cDNA encoding EGF-like

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service