Skip to Content
Merck
All Photos(6)

Key Documents

HPA018846

Sigma-Aldrich

Anti-CSDE1 antibody produced in rabbit

enhanced validation

Prestige Antibodies® Powered by Atlas Antibodies, affinity isolated antibody, buffered aqueous glycerol solution

Synonym(s):

Anti-Cold shock domain-containing protein E1, Anti-N-ras upstream gene protein, Anti-UNR protein

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
12352203
Human Protein Atlas Number:
NACRES:
NA.43

biological source

rabbit

Quality Level

conjugate

unconjugated

antibody form

affinity isolated antibody

antibody product type

primary antibodies

clone

polyclonal

product line

Prestige Antibodies® Powered by Atlas Antibodies

form

buffered aqueous glycerol solution

species reactivity

human

enhanced validation

orthogonal RNAseq
Learn more about Antibody Enhanced Validation

technique(s)

immunoblotting: 0.04-0.4 μg/mL
immunofluorescence: 0.25-2 μg/mL
immunohistochemistry: 1:500-1:1000

immunogen sequence

NLLHNNGHNGYPNGTSAALRETGVIEKLLTSYGFIQCSERQARLFFHCSQYNGNLQDLKVGDDVEFEVSSDRRTGKPIAVKLVKIKQEILPEERMNGQEVFYLTYTPEDVEGNVQLETGDKIN

UniProt accession no.

shipped in

wet ice

storage temp.

−20°C

target post-translational modification

unmodified

Gene Information

human ... CSDE1(7812)

General description

The gene CSDE1 (cold shock domain-containing protein E1) is mapped to human chromosome 1p. CSDE1 is also called as UNR (N-ras upstream gene).

Immunogen

Cold shock domain-containing protein E1 recombinant protein epitope signature tag (PrEST)

Application

All Prestige Antibodies Powered by Atlas Antibodies are developed and validated by the Human Protein Atlas (HPA) project and as a result, are supported by the most extensive characterization in the industry.

The Human Protein Atlas project can be subdivided into three efforts: Human Tissue Atlas, Cancer Atlas, and Human Cell Atlas. The antibodies that have been generated in support of the Tissue and Cancer Atlas projects have been tested by immunohistochemistry against hundreds of normal and disease tissues and through the recent efforts of the Human Cell Atlas project, many have been characterized by immunofluorescence to map the human proteome not only at the tissue level but now at the subcellular level. These images and the collection of this vast data set can be viewed on the Human Protein Atlas (HPA) site by clicking on the Image Gallery link. We also provide Prestige Antibodies® protocols and other useful information.

Biochem/physiol Actions

Cold shock domain-containing protein E1 (CSDE1) is part of the complex that binds c-fos major protein-coding-region determinant of instability (mCRD) and allows interaction between mCRD and the poly(A) tail. This results in RNA deadenylation and decay. CSDE1 particularly have strong affinity for purine-rich sequence. CSDE1 participates in human rhinovirus-2 internal ribosomal entry site (IRES)-mediated translation initiation. GTPase NRAS (transforming protein N-Ras)-CSDE1 is identified as a candidate gene for autism.

Features and Benefits

Prestige Antibodies® are highly characterized and extensively validated antibodies with the added benefit of all available characterization data for each target being accessible via the Human Protein Atlas portal linked just below the product name at the top of this page. The uniqueness and low cross-reactivity of the Prestige Antibodies® to other proteins are due to a thorough selection of antigen regions, affinity purification, and stringent selection. Prestige antigen controls are available for every corresponding Prestige Antibody and can be found in the linkage section.

Every Prestige Antibody is tested in the following ways:
  • IHC tissue array of 44 normal human tissues and 20 of the most common cancer type tissues.
  • Protein array of 364 human recombinant protein fragments.

Linkage

Corresponding Antigen APREST74671

Physical form

Solution in phosphate-buffered saline, pH 7.2, containing 40% glycerol and 0.02% sodium azide

Legal Information

Prestige Antibodies is a registered trademark of Merck KGaA, Darmstadt, Germany

Disclaimer

Unless otherwise stated in our catalog or other company documentation accompanying the product(s), our products are intended for research use only and are not to be used for any other purpose, which includes but is not limited to, unauthorized commercial uses, in vitro diagnostic uses, ex vivo or in vivo therapeutic uses or any type of consumption or application to humans or animals.

Not finding the right product?  

Try our Product Selector Tool.

Storage Class Code

10 - Combustible liquids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

K Xia et al.
Molecular psychiatry, 19(11), 1212-1219 (2013-11-06)
Autism is a highly heritable neurodevelopmental disorder, and known genetic variants, mostly rare, account only for a small proportion of cases. Here we report a genome-wide association study on autism using two Chinese cohorts as gene discovery (n=2150) and three
C Grosset et al.
Cell, 103(1), 29-40 (2000-10-29)
mRNA turnover mediated by the major protein-coding-region determinant of instability (mCRD) of the c-fos proto-oncogene transcript illustrates a functional interplay between mRNA turnover and translation. We show that the function of mCRD depends on its distance from the poly(A) tail.
Frédéric Saltel et al.
Journal of cell science, 130(10), 1796-1808 (2017-04-08)
Unr (officially known as CSDE1) is a cytoplasmic RNA-binding protein with roles in the regulation of mRNA stability and translation. In this study, we identified a novel function for Unr, which acts as a positive regulator of placental development. Unr
Emma C Anderson et al.
The Journal of general virology, 88(Pt 11), 3043-3052 (2007-10-20)
Internal initiation of translation from the human rhinovirus-2 (HRV-2) internal ribosome entry site (IRES) is dependent upon host cell trans-acting factors. The multiple cold shock domain protein Unr and the polypyrimidine tract-binding protein have been identified as synergistic activators of
Elma El Khouri et al.
eLife, 12 (2023-06-21)
A20 haploinsufficiency (HA20) is an autoinflammatory disease caused by heterozygous loss-of-function variations in TNFAIP3, the gene encoding the A20 protein. Diagnosis of HA20 is challenging due to its heterogeneous clinical presentation and the lack of pathognomonic symptoms. While the pathogenic

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service